Skip to Content
Merck
All Photos(2)

Key Documents

MABS417

Sigma-Aldrich

Anti-GOT2, clone 3E9 Antibody

ascites fluid, clone 3E9, from mouse

Synonym(s):

Aspartate aminotransferase, mitochondrial, mAspAT, Fatty acid-binding protein, FABP-1, Glutamate oxaloacetate transaminase 2, Kynurenine aminotransferase 4, Kynurenine aminotransferase IV, Kynurenine-oxoglutarate transaminase 4, Kynurenine--oxoglutarate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$558.00

$558.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$558.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$558.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3E9, monoclonal

species reactivity

mouse, rat, monkey, human

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GOT2(2806)

General description

Aspartate aminotransferase, mitochondrial ((mAspAT), also known as Fatty acid-binding protein (FABP-1) or Glutamate oxaloacetate transaminase 2 or Kynurenine aminotransferase 4 or Kynurenine aminotransferase IV or Kynurenine--oxoglutarate transaminase 4 or Kynurenine--oxoglutarate transaminase IV or Plasma membrane-associated fatty acid-binding protein (FABPpm) or Transaminase A, and encoded by the gene name GOT2, plays an essential role in role in amino acid metabolism and facilitates cellular uptake of long-chain free fatty acids. GOT2 is localized to the mitochondrion; however, exposure to alcohol promotes translocation to the cell membrane.

Immunogen

Purified recombinant fragment of human GOT2 expressed in E. Coli

Application

Anti-GOT2, clone 3E9 Antibody is an antibody against GOT2 for use in western blotting & immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected GOT2 in PC-3 and SK-BR-3 cells.
Research Category
Signaling
Research Sub Category
Metabolic Hormones & Receptors

Quality

Evaluated by Western Blotting in HEK293, PC-12, BCBL-1, HepG2 and NIH/3T3 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected GOT2 in HEK293, PC-12, BCBL-1, HepG2 and NIH/3T3 cell lysates.

Target description

~47 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Xu et al.
Translational stroke research, 11(3), 418-432 (2019-09-02)
The preservation of mitochondrial function is a major protective strategy for cerebral ischemic injuries. Previously, our laboratory demonstrated that protein kinase C epsilon (PKCε) promotes the synthesis of mitochondrial nicotinamide adenine dinucleotide (NAD+). NAD+ along with its reducing equivalent, NADH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service