Skip to Content
Merck
All Photos(1)

Key Documents

MABS385

Sigma-Aldrich

Anti-APOL1 Antibody, clone 1D4

ascites fluid, clone 1D4, from mouse

Synonym(s):

APOL, APO-L, APOL-I, Apolipoprotein L1, Apolipoprotein L, Apolipoprotein L-I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$744.00

$744.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$744.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$744.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1D4, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APOL1(8542)

General description

Apolipoprotein L1/Apolipoprotein L-1 (ApoL-1), or alternatively Apolipoprotein L (Apo-L/ApoL) is the protein encoded by the APOL1 gene in humans. Apolipoprotein L1 is one of the major carriers of cholesterol in the blood. It plays a role in the transport of cholesterol from the peripheral cells to the liver. It is a secreted protein expressed in liver, pancreas, lung, prostate, placenta and spleen. Apolipoprotein L1 is part of the HDL (high density lipoprotein) complex and Apolipoprotein L1 possesses both extra- and intra-cellular functions crucial in host defense and cellular homeostatic mechanisms. Alterations in Apolipoprotein L1 function due to genetic, environmental, and lifestyle factors have been associated with African sleeping sickness, atherosclerosis, lipid disorders, obesity, schizophrenia, cancer, and chronic kidney disease (CKD), yet much of its biological function remains unknown. Interesting, Apolipoprotein L1 has the ability to kill circulating trypanosomes in the blood. Trypanosomes cause a serious disease called sleeping sickness but in those individuals that have particular splice variants of Apolipoprotein L1 are spared sleeping sickness, however the variants are associated with an increased chance of kidney disease as a tradeoff. Apolipoprotein L1 appears important in autophagy and even as a cancer preventative as well. Recent research shows that increased intracellular levels of Apolipoprotein L1 can induce autophagy and autophagy-associated cell death but that Apolipoprotein L1 may protect kidney cells against renal cell carcinoma.

Immunogen

Purified recombinant fragment of human APOL1 expressed in E. Coli.

Application

Detect Apolipoprotein using this mouse monoclonal antibody, Anti-APOL1 Antibody & clone 1D4 validated for use in western blotting.

Quality

Evaluated by Western Blotting in human plasma.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected APOL1 in human plasma.

Optimal working dilutions must be determined by end user.

Target description

~44 kDa observed

Analysis Note

Control
Human plasma lysate

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service