Skip to Content
Merck
All Photos(1)

Key Documents

MABS351

Sigma-Aldrich

Anti-FAT10 Antibody, clone 4F1

clone 4F1, from mouse

Synonym(s):

Ubiquitin D, Diubiquitin, Ubiquitin-like protein FAT10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$660.00


Estimated to ship on11 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F1, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBD(10537)

General description

FAT10 protein is a recombinant protein produced in E. coli corresponding to the human sequence encompassing an N-terminal His6-tag within a leader sequence. FAT10, a ubiquitin-like protein, is encoded in the major histocompatibility complex class I locus and is synergistically inducible with interferon-gamma and tumor necrosis factor alpha. FAT10 expression causes apoptosis and it appears that FAT10 may modulate tumorigenesis through its interaction with the MAD2 spindle-assembly checkpoint protein. It has been demonstrated that FAT10 and its conjugates are rapidly degraded by the proteasome. Furthermore, a new interaction partner of FAT10, NEDD8 ultimate buster-1L (NUB1L), may function as a linker that targets FAT10 for degradation by the proteasome.

Immunogen

GST-tagged recombinant protein corresponding to human FAT10.

Application

Anti-FAT10 Antibody, clone 4F1 is a highly specific mouse monoclonal antibody, that targets FAT10 & has been tested in western blotting, IP & ICC.
Immunocytochemistry Analysis: A representative lot detected FAT10 in HA-FAT10 transfected HEK293 cells (Aichem, A., et al. (2010). Nature Commun. 1:13).
Immunoprecipitation Analysis: A representative lot immunoprecipitated FAT10 in transfected HA-FAT10 HEK293 cells cultivated with IFNgamma and TNFalpha (Aichem, A., et al. (2010). Nature Commun. 1:13).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected FAT10 in 10 µg of HepG2 cell lysate.

Target description

~18 kDa observed. FAT10 FAT10ylates itself as well as other proteins so additional bands may be observed due to the FAT10ylation of endogenous proteins (Aichem, A., et al. (2010). Nature Commun. 1:13.),(Aichem, A., et al. J Cell Sci. (2012)125:4576-85).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenming Zhang et al.
American journal of cancer research, 14(4), 1523-1544 (2024-05-10)
Although sorafenib is the first-line therapeutic agent for advanced hepatocellular carcinoma (HCC), the development of drug resistance in HCC cells limits its clinical efficacy. However, the key factors involved in mediating the sorafenib resistance of HCC cells and the underlying

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service