Skip to Content
Merck
All Photos(1)

Key Documents

MABS278

Sigma-Aldrich

Anti-EMAPII Antibody, clone M7/1, (Preservat ive free)

clone M7/1, from rat

Synonym(s):

multisynthetase complex auxiliary component p43, multisynthetase complex auxiliary component 1, ARS-interacting multifunctional protein 1, Aminoacyl tRNA synthase complex-interacting multifunctional protein, Multisynthase complex auxiliary component p43,

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$578.00

$578.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$578.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$578.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M7/1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (immunogen homology)

technique(s)

activity assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AIMP1(9255)

General description

Endothelial monocyte-activating polypeptide II (EMAPII) is a 22 kDa pro-inflammatory cytokine that regulates TNFβ signaling. The mature EMAPII is formed from a functional EMAPII precursor protein in response to apoptosis, and other forms of cell stress. Previous studies have reported that it may decrease the growth of tumors by inhibiting angiogenesis. EMAPII has also been implicated in autoimmune diseases. However, EMAPII is also involved in a range of normal biological processes. It is a component of the multisynthase complex that binds tRNA and plays an essential role in protein translation, and may also play a role in lung development.

Specificity

This antibody recognizes the mature form of this protein at ~23 kDa and the larger pro EMAP II form at ~43 kDa which is secreted based on lysate.

Immunogen

His-tagged recombinant protein corresponding to mouse EMAPII.

Application

Anti-EMAPII Antibody, clone M7/1, (Preservat ive free) detects level of EMAPII & has been published & validated for use in Western Blotting, Activity Assay & IHC.
Research Category
Signaling
Research Sub Category
Developmental Signaling
Western Blot Analysis: A representative lot was used by an independent laboratory in recombinant EMAP II proteins. (Rajashekhar, G., et al. (2009). J Immunol Methods. 350(1-2):22–28.)

Neutralization Antibody: A representative lot was used by an independent laboratory to functionally neutralize pro and mature forms of EMAPII in lung tissue lysate. (Clauss, M., et al. (2012). J Clin Invest doi:10.1172/JCI43881.)

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in lung parenchyma of patients with emphysema. (Clauss, M., et al. (2012). J Clin Invest doi:10.1172/JCI43881.)

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected EMAPII in 10 µg of HeLa cell lysate.

Target description

~43 kDa observed. This antibody recognizes the mature form of this protein at ~23 kDa and the larger pro EMAP II form at ~43 kDa which is secreted based on lysate.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service