Skip to Content
Merck
All Photos(2)

Key Documents

MABS270

Sigma-Aldrich

Anti-PP2A reg. subunit B56 epsilon (PPP2R5E) Antibody, clone 5A5-1F3

clone 5A5-1F3, from mouse

Synonym(s):

Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform, PP2A B subunit isoform B′-epsilon,

PP2A B subunit isoform B56-epsilon, PP2A B subunit isoform PR61-epsilon, PP2A B subunit isoform R5-epsilon

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$750.00

$750.00


Estimated to ship on10 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$750.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$750.00


Estimated to ship on10 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A5-1F3, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPA(5524)

General description

Protein phosphatase 2A (PP2A) is a family of ubiquitous heterotrimeric proteins that catalyze the removal of phosphate groups from serine and threonine residues. The PP2A enzymes contain core structural (A) and catalytic (C) subunits and variable regulatory B subunits. The B subunits regulate their catalytic activity, as well as dictate substrate specificity and subcellular localization. Recent studies have suggested that B56 epsilon subunits regulate p53-mediated apoptotic pathways, and can induce or suppress apoptosis depending on the cellular context. PP2A enzymes are also regulated by posttranslational modifications and endogenous inhibitors.

Specificity

This antibody is specific for the B56 regulatory subunit of Protein phosphatase 2A (PP2A).

Immunogen

His-tagged recombinant protein corresponding to human PP2A reg. subunit B56 epsilon (PPP2R5E).

Application

Research Category
Neuroscience

Apoptosis & Cancer
Research Sub Category
Kinases & Phosphatases

Apoptosis - Additional
This Anti-PP2A reg. subunit B56 epsilon (PPP2R5E) Antibody, clone 5A5-1F3 is validated for use in Western Blotting, ICC, Immunoprecipitation for the detection of PP2A reg. subunit B56 epsilon (PPP2R5E).
Western Blot Analysis: A 1:1,000 dilution from a representative lot detected PP2A reg. subunit B56 epsilon (PPP2R5E) in 10 µg of U2OS cell lysate and mouse brain tissue lysate.

Immunocytochemistry Analysis: A representative lot detected PP2A reg. subunit B56 epsilon (PPP2R5E) in U2OS cells. (Max F. Perutz Laboratories, Medical University of Vienna, Austria).

Immunoprecipitation Analysis: A representative lot immunoprecipitated endogenous PP2A reg. subunit B56 epsilon (PPP2R5E) in a panel of U2OS cell lysates expressing the B56 alpha, beta, gamma, delta, and epsilon regulatory subunit of PP2A (Max F. Perutz Laboratories, Medical University of Vienna, Austria).

Quality

Evaluted by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: A 1:1,000 dilution from a representative lot detected PP2A reg. subunit B56 epsilon (PPP2R5E) in 10 µg of NIH/3T3 cell lysate.

Target description

~50 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrich Baumgartner et al.
Molecular cancer, 17(1), 44-44 (2018-02-20)
Epidermal growth factor receptor (EGFR) mutations enable constitutive active downstream signaling of PI3K/AKT, KRAS/ERK and JAK/STAT pathways, and promote tumor progression by inducing uncontrolled proliferation, evasion of apoptosis and migration of non-small cell lung cancer (NSCLC). In addition, such EGFR
Huan He et al.
eLife, 7 (2018-10-16)
The intracellular parasite Toxoplasma gondii, hijacks evolutionarily conserved host processes by delivering effector proteins into the host cell that shift gene expression in a timely fashion. We identified a parasite dense granule protein as GRA18 that once released in the
Qing Kang et al.
The Journal of biological chemistry, 289(25), 17480-17496 (2014-05-16)
The mitotic kinase Aurora A (AurA) is regulated by a complex network of factors that includes co-activator binding, autophosphorylation, and dephosphorylation. Dephosphorylation of AurA by PP2A (human, Ser-51; Xenopus, Ser-53) destabilizes the protein, whereas mitotic dephosphorylation of its T-loop (human
Jade J Dyson et al.
FASEB bioAdvances, 4(4), 273-282 (2022-04-14)
Protein phosphatase 2A (PP2A) is a heterotrimeric phosphatase that controls a wide range of cellular functions. The catalytic activity and intracellular location of PP2A are modulated by its association with regulatory B subunits, including B56 proteins, which are encoded by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service