Skip to Content
Merck
All Photos(1)

Key Documents

MABS2278

Sigma-Aldrich

Anti-Oxidized phospholipid Antibody, clone EO6

Synonym(s):

OX-Phospholipid, Ox-PC

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$233.00
100 μL
$590.00

$233.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$233.00
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$233.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

E06, monoclonal

mol wt

calculated mol wt N/A kDa
observed mol wt ~200 kDa

purified by

affinity chromatography

species reactivity

all

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgMκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Specificity

Clone E06 is a mouse monoclonal antibody that binds to oxidized phospholipids and reacts with oxidized LDL, oxidized HDL and proteins covalently modified by oxidized phospholipids.

Immunogen

B lymphocytes from the spleens of ApoE-deficient mice fused with a myeloma cell line and selected hybridomas were cloned and cells were injected intraperitoneally into mice as described by Hörkkö et al. (1999).

Application

Quality Control Testing

Evaluated by ELISA with Oxidized Phosphatidylcholine-BSA (OxPC-BSA)

ELISA Analysis: A serial dilution of this antibody detected Oxidized-Phosphatidylcholine-BSA.

Tested Applications

Western Blotting Analysis: A representative lot detected OxPL in Western Blotting applications (Horkko, S., et al. (1999). J Clin Invest. 103(1):117-28; Leibundgut, G., et al. (2013). J Lipid Res. 54(10)2815-30).

ELISA Analysis: A representative lot detected OxPL in ELISA applications (Horkko, S., et al. (1999). J Clin Invest. 103(1):117-28).

Immunohistochemistry Applications: A representative lot detected Oxidized phospholipid in Immunohistochemistry applications (Yu, P., et al. (2018). Arterioscler Thromb Vasc Biol. 38(1):26-39).

Radioimmunoassay: A representative lot detected OxPL in Radioimmunoassay applications (Horkko, S., et al. (1999). J Clin Invest. 103(1):117-28).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Several reactive lipid peroxidation products are generated during the oxidation of LDL, and modification of lipids and apoproteins occurs in vivo. These oxidized and/modified LDL have the ability to bind to scavenger receptors that can lead to unregulated accumulation of cholesterol in macrophages and formation of foam cells. These products can also react with lipids, apoproteins, and proteins and generate immunogenic neoepitopes and contribute to atherogenesis. The lipid peroxidation products can for adducts with free amino groups of lysines and other amino acids in apolipoproteins. Reactive oxygen species (ROS) and oxidized phospholipids are also reported to activate TRPA1 and TRPV1 cation ion channels that shape thermal and mechanical hypersensitivity in inflammation and pain. These channels are also suggested to be involved in pathological events, including rheumatoid arthritis and trigeminal neuropathic pain. Even locally injected oxidized lipids can elicit thermal and mechanical hypersensitivity and acute nociception. Clone E06/T15 is a monoclonal autoantibody that specifically binds to the head group of oxidized phosphatidylcholines in oxidized phospholipids. (Ref.: Mohammadi, M., et al. (2018). Br. J. Pharmacol. 175(14); 2940-2955; Hörkkö, S., et al. (1999). J. Clin. Invest. 103(1): 117-128; Palinski, W., et al. (1996). J. Clin. Invest. 98:800 814.).

Physical form

Purified mouse monoclonal antibody IgM in PBS without preservatives.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service