Skip to Content
Merck
All Photos(1)

Key Documents

MABS2066

Sigma-Aldrich

Anti-LLGL1 Antibody, clone mAb17-35

clone mAb17-35, from mouse

Synonym(s):

Lethal(2) giant larvae protein homolog 1, Mgl-1, Mlgl

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$252.00
100 μG
$666.00

$252.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$252.00
100 μG
$666.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAb17-35, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Llgl1(16897)

General description

Lethal(2) giant larvae protein homolog 1 (UniProt: Q80Y17; also known as LLGL, Mgl-1, Mlgl) is encoded by the Llgl1 (also known as Llglh) gene (Gene ID: 16897) in murine species. LLGL belongs to the WD repeat L(2)GL family. LLGL is a cortical cytoskeleton protein that is found in a complex involved in maintenance of cell polarity and epithelial integrity. It is involved in the regulation of mitotic spindle orientation, proliferation, differentiation, and tissue organization of neuroepithelial cells. It is generally enriched in developing axons. LLGL stability and function is achieve by its interaction with RanBPM protein, which primarily acts as a scaffolding protein. Its N-terminal region containing SPRY domain is shown to be responsible for binding to RanBPM and this binding functionally extends its half-life by preventing its turnover through ubiquitin-proteasomal pathway. RanBPM is shown enhance tumor suppressor activity of LLGL by inhibiting LLGL-mediated cell proliferation and cell migration. (Ref.: Suresh, B., et al. (2010). J. Biol. Chem. 285(46); 35340-35349).

Specificity

Clone mAb17-35 specifically detects LLGL1 in mouse. It targets an epitope with in 153 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
GST-tagged recombinant fragment corresponding to 153 amino acids from the C-terminal region of murine Lethal(2) giant larvae protein homolog 1 (LLGL).

Application

Anti-LLGL1, clone mAb17-35, Cat. No. MABS2066, is a mouse monoclonal antibody that detects Lethal(2) giant larvae protein homolog 1 and has been tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: A representative lot detected LLGL1 in Western Blotting applications (Ivanov, A.I., et. al. (2010). Am J Pathol. 176(1):134-45).

Immunocytochemistry Analysis: A representative lot detected LLGL1 in Immunocytochemistry applications (Ivanov, A.I., et. al. (2010). Am J Pathol. 176(1):134-45).

Immunohistochemistry Analysis: A representative lot detected LLGL1 in Immunohistochemistry applications (Weimer, J.M., et. al. (2009). Development. 136(17):2965-75).

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected LLGL1 in mouse brain tissue lysate.

Target description

~120 kDa observed; 112.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service