Skip to Content
Merck
All Photos(2)

Key Documents

MABS2036M

Sigma-Aldrich

Anti-Cytochrome b Antibody, clone 5B3-6E3

clone 5B3-6E3, from mouse

Synonym(s):

Cytochrome b (UniProt: P00156, also known as Complex III subunit 3, Complex III subunit III, Cytochrome b-c1 complex subunit 3, Ubiquinol-cytochrome-c reductase complex cytochrome b subunit

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00
100 μG
$512.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$214.00
100 μG
$512.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5B3-6E3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MT-CYB(4519)

General description

Cytochrome b (UniProt: P00156; also known as Complex III subunit 3, Complex III subunit III, Cytochrome b-c1 complex subunit 3, Ubiquinol-cytochrome-c reductase complex cytochrome b subunit) is encoded by the MT-CYB (also known as COB, CYTB, MTCYB) gene (Gene ID: 4519) in human. Cytochrome b is a mitochondrial inner membrane protein that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex). It is a part of the mitochondrial respiratory chain. The b-c1 complex mediates electron transfer from ubiquinol to cytochrome c and contributes to the generation of a proton gradient across the mitochondrial membrane, which is used for ATP synthesis. The MT-CYB gene is a good phylogenetic marker. Defects in MT-CYB gene are a rare cause of mitochondrial dysfunction underlying different myopathies, which include mitochondrial encephalomyopathy, hypertrophic cardiomyopathy, and sporadic mitochondrial myopathy. Some cases of Leber hereditary optic neuropathy (LHON) are also linked to defects in MT-CYB gene, which result in acute or subacute loss of central vision, due to optic nerve dysfunction.

Specificity

Clone 5B3-6E3 detects cytochrome b in human mitochondria. It targets an epitope within the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated Synthetic peptide corresponding to the N-terminus of human cytochrome b.

Application

Anti-Cytochrome b, clone 5B3-6E3, Cat. No. MABS2036, is a mouse monoclonal antibody that detects Complex III Cytochrome b and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 0.1 µg/mL from a representative lot detected Cytochrome b in mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA. (Courtesy of Michael F. Marusich, Ph.D., mAbDx, Inc., Eugene, OR USA)

Quality

Evaluated by Western Blotting in Mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected Cytochrome b in Mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA.

Target description

~28 kDa observed; 42.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein L
Purified mouse monoclonal antibody IgG2b in buffer containing HEPES-Buffered Saline (150 mM NaCl, 15 mM HEPES, pH 7.2) with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Sheng Zhang et al.
Nature cell biology, 23(7), 684-691 (2021-07-14)
Members of the mammalian AlkB family are known to mediate nucleic acid demethylation1,2. ALKBH7, a mammalian AlkB homologue, localizes in mitochondria and affects metabolism3, but its function and mechanism of action are unknown. Here we report an approach to site-specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service