Skip to Content
Merck
All Photos(2)

Key Documents

MABN863

Sigma-Aldrich

Anti-Cav1.2 alpha1c, clone 8F9.1 Antibody

clone 8F9.1, from mouse

Synonym(s):

Voltage-dependent L-type calcium channel subunit alpha-1C, Calcium channel, L type. alpha-1 polypeptide, isoform 1, cardiac muscle, Voltage-gated calcium channel subunit alpha Cav1.2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Estimated to ship on16 April 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1293

Request a Bulk Order

Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$589.00


Estimated to ship on16 April 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1293

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8F9.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CACNA1C(775)

General description

This gene encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by this gene binds to and is inhibited by dihydropyridine. Many alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.

Immunogen

Recombinant protein corresponding to human Cav1.2 alpha1c.

Application

Anti-Cav1.2 alpha1c, clone 8F9.1 | MABN863 is an antibody against Cav1.2 alpha1c for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected Cav1.2 alpha1c in human cerebellum, human cerebral cortex, and human pancreas tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 5.0 µg/mL of this antibody detected Cav1.2 alpha1c in 10 µg of human brain tissue lysate.

Target description

~230 kDa observed. UniProt entry Q13936 describes 37 isoforms produced by alternative splicing.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service