Skip to Content
Merck
All Photos(2)

Key Documents

MABN841

Sigma-Aldrich

Anti-Semaphorin 4A Antibody, clone 8B5.1

clone 8B5.1, from mouse

Synonym(s):

Semaphorin 4A, Semaphorin-B, Sema B, Sema4A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$680.00

$680.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$680.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$680.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8B5.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SEMA4A(64218)

General description

Semaphorin-4A (UniProt Q9H3S1; also known as Sema B, Semaphorin-B) is encoded by the SEMA4A (also known as CORD10, RP35, SEMAB, SEMB) gene (ORP UNQ783/PRO1317; Gene ID 64218) in human. Semaphorin-4A is a membrane-type class IV semaphorin originally characterized for its role in axon guidance during neural development, Semaphorin-4A also plays a role in Th cells activation and the differentiation of Th1 and Th17 cells. In addition Semaphorin-4A is involved in angiogenesis and endothelial cells migration by modulating VEGF signaling. Consistent with its important role in Th cells, Semaphorin-4A level is reported to be elevated in the serum samples from patients with multiple sclerosis (MS) respond poorly to interferon-beta (IFN-β) therapy. Semaphorin-4A neutralizing antibodies are reported to inhibit antigen-specific T cell generation and ameliorate experimental autoimmune encephalomyelitis (EAE), an animal model of MS.

Specificity

Expected to react with both spliced isoforms.

Immunogen

Epitope: Sema domain
GST-tagged recombinant protein corresponding to the Sema domain of human Semaphorin 4A.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Semaphorin 4A Antibody, clone 8B5.1 is validated for use in Western Blotting for the detection of Semaphorin 4A.
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected Semaphorin 4A in 10 µg of human retinal pigment epithelia (RPE) ARPE-19.

Quality

Evaluated by Western Blotting in human monocytic leukemia THP-1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Semaphorin 4A in 10 µg of human monocytic leukemia THP-1 cell lysate.

Target description

~85 kDa observed. 83.6 kDa (isoform 1) and 69.1 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service