Skip to Content
Merck
All Photos(3)

Key Documents

MABN804

Sigma-Aldrich

Anti-Ribeye Antibody, clone 3D12.1

clone 3D12.1, from mouse

Synonym(s):

C-terminal-binding protein 2, CtBP2, Ribeye

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$680.00

$680.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$680.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$680.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D12.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CTBP2(1488)

General description

Human C-terminal-binding protein 2 (CtBP2) and Ribeye (UniProt P56545-1 and P56545-2, respectively) are two proteins encoded by the CTBP2 gene (Gene ID 1488) in human, they are produced as a result of alternative splicing. The C-terminal 425-amino acid sequence of Ribeye (aa 561-985) is identical to that of CtBP2 (aa 21-445), however the first 20 amino acids at the CtBP2 N-terminal end is replaced with a 560-amino acid sequence in the case of Ribeye. Ribeye is a protein specific to synaptic ribbons. Ribbon synapses are high-performance synapses built by certain nonspiking sensory neurons, such as photoreceptors, retinal bipolar cells, and inner ear hair cells. The main site of exocytosis of these neurons is characterized by unique presynaptic structures called the synaptic ribbons that associate with large numbers of synaptic vesicles primed for exocytosis and has a direct impact on synaptic signaling. Ribeye is composed of an N-terminal A domain and a C-terminal B domain that is largely identical to CtBP2. The A domain unique to Ribeye has a structural role at the synaptic ribbon, while the B domain of Ribeye possesses lysophosphatidic acid (LPA) acyltransferase (LPAAT) activity that catalyzes the generation of the highly active signaling phospholipid phosphatidic acid (PA) from LPA. LPAAT-deficient Ribeye mutant does not recruit PA-binding proteins to synaptic ribbons, whereas wild-type RIBEYE supports PA-dependent binding, The B domain also contains an NADH-binding subdomain (NBD), which is reported to compete against Arf1 for Arf-GTPase activating protein-3 (ArfGAP3) binding. Ribeye therefore functions as an Arf1 positive regulator at the synaptic ribbon by preventing the inactivation of Arf1 by ArfGAP3.

Specificity

Specific for CTBP2 spliced variant Ribeye. Does not react with the canonical CtBP2 isoform.

Immunogen

Epitope: N-terminal region
GST-tagged recombinant protein corresponding to the N-terminal region of human Ribeye.

Application

Anti-Ribeye Antibody, clone 3D12.1, Cat. No. MABN804, is a highly specific mouse monoclonal antibody, that targets Ribeye and has been tested in Western Blotting and Immunohistochemistry (Paraffin).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Ribeye in 10 µg of SH-SY5Y cell lysate.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected Ribeye in human cardiac myocytes and human cerebellum tissues.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Ribeye in 10 µg of HeLa cell lysate.

Target description

~106 kDa observed (109.2 kDa calculated).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service