Skip to Content
Merck
All Photos(1)

Key Documents

MABN768

Sigma-Aldrich

Anti-PrP, clone DRM2-118 Antibody

clone DRM2-118, from mouse

Synonym(s):

Major prion protein, PrP, PrP27-30, PrP33-35C, CD230

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$589.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DRM2-118, monoclonal

species reactivity

rat, bovine, mouse, porcine, hamster, sheep, human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRNP(5621)

General description

The protein named PrP is the cellular gene for the prion protein. The normal form appears to play a role in neuronal development and synaptic plasticity, iron transport and myelin sheath maintenance. PrP has both membrane associated and cytoplasmic/nuclear isoforms. The primary isoform is localized to the cellular membrane in lipid anchor clusters, as well as in the Golgi. Isoform 2 is cytoplasmic that becomes localized to the nucleus. Prp is normally a monomeric protein but a disease associated mutated form will form protease resistant oligomers often with another protein beta amyloid. The mutated form is found in high quantity in the brain of humans and animals infected with the degenerative neurological diseases such as Kuru, Creutzfeldt-Jakob disease (CJD), Gerstmann-Straussler syndrome (GSS), Scrapie, and Bovine Spongiform Encephalopathy (BSE) and Transmissible Mink Encephalopathy.

Specificity

DRM1-60 showed strong binding to only one peptide (#29: NQVYYRPVDQYS) suggesting a linear epitope corresponding to SHaPrP(159–170), located in b2-a2 loop. This epitope assignment is consistent with the binding of DRM1-60 to PrPC to all species tested, except human and horse, which both harbor substitutions in this region. While antibodies with epitopes similar to DRM2-118 have been described,(36) DRM1-60 and DRM1-31 have unique epitopes not previously reported. DRM1-60 appears to bind a linear epitope localized to the b2-a2 loop, while DRM1-31 has a conformational epitope located on the exposed surface formed by the close proximity of the b2-a2- helix loop, the N-terminus of the a2 helix, and the C-terminus of the a3 helix.

Immunogen

Recombinant protein corresponding to syrian hamster PrP.

Application

Anti-PrP, clone DRM2-118 | MABN768 is an antibody against PrP for use in Western Blotting, Immunohistochemistry, ELISA.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected PrP in 10 µg of rat brain tissue lysate.
ELISA Analysis: A representative lot detected PrP in recombinant PrP proteins from multiple species by ELISA using DRM1-31, DRM1-60, and DRM2-118 (Stanker, L.H., et al. (2012). 31(5):314-24).
Immunohistochemistry Analysis: A representative lot detected PrP in hamster brain sections tissue with DRM1-31, DRM1-60, and DRM2-118 (Stanker, L.H., et al. (2012). 31(5):314-24).
Western Blotting Analysis: A representative lot detected PrP in normal brain extract from multiple spcecies using DRM2-118. DRM1-60, and DRM1-31 lysate (Stanker, L.H., et al. (2012). 31(5):314-24).

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PrP in 10 µg of mouse brain tissue lysate.

Target description

~36 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service