Skip to Content
Merck
All Photos(1)

Key Documents

MABN757

Sigma-Aldrich

Anti-Beta-glucosidase 2 (GBA2) Antibody, clone 4A12

clone 4A12, from rat

Synonym(s):

Non-lysosomal glucosylceramidase, Beta-glucocerebrosidase 2, Beta-glucosidase 2, Glucosylceramidase 2, NLGase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A12, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Gba2(230101)

General description

Non-lysosomal glucosylceramidase (EC 3.2.1.45; UniProt Q69ZF3; also known as Beta-glucocerebrosidase 2, Beta-glucosidase 2, Glucosylceramidase 2, NLGase) is encoded by the Gba2 (also known as Kiaa1605) gene (Gene ID 230101) in murine species. Glucosidases GBA1, GBA2, and GBA3 are structurally unrelated proteins with similar hydrolase activity that mediates glucosylceramide (GlcCer) degradation. GBA1 mediates GlcCer degradation in the lysosome lumen following its transport via the endosomal pathway. GBA2 is localized at the cytosolic surface of the ER and Golgi, with a predominant presence at the cis-Golgi in close proximity to the site of GlcCer synthesis and sphingomyelin conversion. GlcCer is also delivered to the ER through FAPP2-dependent transport and then flips to ER lumenal side by low specificity phospholipid flippases. GBA2 ER localization allows it to regulate GlcCer level at the cytosolic side of the ER and limit the amount of GlcCer from entering the ER lumenal side for higher order glycosphingolipid synthesis. Impaired GBA1 activity causes accumulation of GlcCer in lysosomes of tissue macrophages, causing liver and spleen enlargement and impairment of the central nervous system in patients with Gaucher disease. GBA2 activity is found downregulated in fibroblasts from a patient with type II Gaucher disease and fibroblasts from Gba1-deficient mice, suggesting a cross-talk between GBA1 and GBA2 that could affect the phenotype of Gaucher disease. GBA3 is a cytosolic Klotho-related protein whose physiological function is not yet known.

Specificity

Clone 4A12 detected the beta-glucosidase 2 (GBA2) target band in testis tissue sample from wild-type, but not Gba2-knock mice (Körschen, H.G., et al. (2013). J. Biol. Chem. 288(5):3381-3393).

Immunogen

Ovalbumin-conjugated linear peptide corresponding to a sequence from the N-terminal region of mouse beta-glucosidase 2 (GBA2).

Application

Anti-Beta-glucosidase 2 (GBA2) Antibody, clone 4A12, Cat. No. MABN757, is a highly specific rat monoclonal antibody that targets beta-glucosidase 2 (GBA2) and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A representative lot immunostained 4% paraformaldehyde-fixed, 0.5% Triton X-100-permeabilized, but not non-permeabilized, HEK293 transfectants expressing HA-tagged mouse beta-glucosidase 2 (GBA2-HA). GBA2-HA immunoreactivity was found associated with both Golgi and ER markers and greatly dependent on microtubule dynamics (Körschen, H.G., et al. (2013). J. Biol. Chem. 288(5):3381-3393).

Immunocytochemistry Analysis: A representative lot detected beta-glucosidase 2 (GBA2) in isolated embryonic mouse hippocampal neurons, but not astrocytes (Körschen, H.G., et al. (2013). J. Biol. Chem. 288(5):3381-3393).

Western Blotting Analysis: A representative lot detected HA-tagged mouse beta-glucosidase 2 exogenously expressed in HEK293 cells, as well endogenous beta-glucosidase 2 in testis tissue samples from wild-type, but not Gba2-knock mice (Körschen, H.G., et al. (2013). J. Biol. Chem. 288(5):3381-3393).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in mouse brain tissue sections.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected beta-glucosidase 2 (GBA2) in mouse brain tissue sections.

Target description

103.3 kDa calculated.

Physical form

Format: Purified
Protein G purified.
Purified rat IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service