Skip to Content
Merck
All Photos(1)

Key Documents

MABN748

Sigma-Aldrich

Anti-TrpC3, clone 10H6 Antibody

clone 10H6, from mouse

Synonym(s):

Short transient receptor potential channel 3, TrpC3, Transient receptor protein 3, TRP-3, hTrp-3, hTrp3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$524.00

$524.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$524.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$524.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10H6, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRPC3(7222)

General description

TrpC3, also known as Short transient receptor potential channel 3, transient receptor protein 3 or TRP-3, and encoded by the gene TRPC3/TRP3, forms a receptor activated calcium channel that is activated via ligand binding and transduces signals via the G-protein coupled tyrosine receptor kinases pathways.TrpC3 can also be activated by the release of internal calcium and is one of the principal receptors involved in maintaining Ca++ ion homeostasis in skeletal muscles and nervous tissue. TrpC3 is also a principal receptor involved in the Capacitative calcium entry (CCE) which is the mechanism behind Ca++ influx into cells that replenishes internal calcium stores. Additionally, TrpC3 receptor is important in axon guidance, phototransduction, and platelet activation. TrpC3 is localized to the cell membrane and consists of a multipass-though transport protein. TrpC3 is expressed principally in the brain but is also found in most other tissues and organs at lowered levels.

Immunogen

Recombinant protein corresponding to mouse TrpC3.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-TrpC3, clone 10H6 Antibody is validated for use in Western Blotting for the detection of TrpC3.
Western Blotting Analysis: A representative lot detected TrpC3 in cerebellar tissues of TRPC3 wild type mice and not TRPC3 knock out mice (Prof. Yizheng Wang, Shanghai Institutes for Biological Sciences (SIBS).
Western Blotting Analysis: A representative lot detected TrpC3 in rat liver and brain tissue lysates (Feng. S., et al. (2013). PNAS. 1-6).

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected TrpC3 in 10 µg of mouse brain tissue lysate.

Target description

~110 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service