Skip to Content
Merck
All Photos(3)

Key Documents

MABN674

Sigma-Aldrich

Anti-PSIP1 Antibody, clone 6E4

ascites fluid, clone 6E4, from mouse

Synonym(s):

PSIP1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$584.00

$584.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$584.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$584.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6E4, monoclonal

species reactivity

human, rat, monkey

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PSIP1(11168)

General description

PSIP1, also known as coactivator p75, as well as numerous other synonyms named after their original discoveries including, PC4 and SFRS1 interacting protein 1, lens epithelium-derived growth factor (LEDGF/p75), dense fine speckles 70kD protein (DFS 70), and transcriptional coactivator p75/p52, is a transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. It is particularly important in lens epithelial cell gene regulation and stress responses, and even though initially thought to be limited to the lens of the eye, PSIP1 is now known to be widely expressed in most tissues and has no direct role in the development of lens epithelium. It also gained research interest as the focal point for HIV integration and appears to be one of the few integrase interactor proteins whose knockdown limits HIV integration and propagation. Moreover, PSIP1 is an anti-apoptotic protein and appears to play a protective role during stress-induced apoptosis. There are three isoforms of the PSIP1 in humans, PSIP1, PSIP2 or p52, and a third isoform which is unnamed. The PSIP1 isoforms seem to vary in their precise functions somewhat with isoform 2 being the most commonly expressed and most abundant. Expression is localized to the nucleus. EMD-Millipore’s anti-PSIP1mouse monoclonal, clone 6.00E+04 has been tested on human, paraffin embedded immunohistochemical sections, immunocytochemical immunofluorescence studies using mouse NIH3T3 cells, western blot using human, mouse, and rat cells line lysates, and ELISA using purified antigen.

Immunogen

Purified recombinant fragment of human PSIP1 expressed in E. Coli.

Application

Detect PSIP1 using this mouse monoclonal antibody, Anti-PSIP1 Antibody, clone 6E4 validated for use in western blotting, IHC & Immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected PSIP1 in NIH/3T3 cells.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected PSIP1 in rat cerebellum, rat hippocampus, and human cerebellum tissues.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in HepG2, Jurkat, K562, Cos7, PC-12, HeLa, and NIH/3T3 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected PSIP1 in HepG2, Jurkat, K562, Cos7, PC-12, HeLa, and NIH/3T3 cell lysates.

Target description

~75 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HepG2, Jurkat, K562, Cos7, PC-12, HeLa, and NIH/3T3 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiffany Sin Yu Chan et al.
Cancer research, 76(9), 2802-2812 (2016-03-26)
Substantial evidence links Myc-PI3K/AKT signaling to the most aggressive subtype of medulloblastoma and this axis in medulloblastoma therapy. In this study, we advance understanding of how Myc-PI3K/AKT signaling contributes to this malignancy, specifically, in identifying the Myc-interacting protein JPO2 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service