Skip to Content
Merck
All Photos(2)

Key Documents

MABN673

Sigma-Aldrich

Anti-ARNTL Antibody, clone 1C5

ascites fluid, clone 1C5, from mouse

Synonym(s):

Aryl hydrocarbon receptor nuclear translocator-like protein 1, Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Short name=bHLHe5, Member of PAS protein 3, PAS domain-containing protein 3, b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$545.00

$545.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$545.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$545.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1C5, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARNTL(406)

General description

Aryl hydrocarbon receptor nuclear translocator-like protein 1 or alternatively, Basic-helix-loop-helix PAS protein MOP3, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5 (bHLHe5), Member of PAS protein 3, PAS domain-containing protein 3, bHLH-PAS protein JAP3, and encoded by the human gene ARNTL,BHLHE5, BMAL1, MOP3 or PASD3, is a protein belonging to the basic helix-loop-helix-PAS domain containing transcription factor family. ARNTL protein is important for the molecular mechanisms behind circadian rhythms as it interacts with a number of CLOCK (Circadian Locomotor Output Cycles Kaput) family members as well as CLOCK itself to promote transcription of a number of proteins of the circadian clock. ARNTL interacts with CRY and CRY2, PER, TIMELESS, and other clock related genes as well as non-CLOCK proteins such as HIF-1alpha (HIF1A) and SUMO3. ARNTL appears critical for proper function of rhythms, as only knockouts of ARNTL in mice generate mice that have upset biological clocks and a shorter lifespan. ARNTL is highly expressed in the adult brain as well as skeletal muscle and heart and ARNTL is a nuclear localized transcription activator. EMD-Millipore’s Anti-ARNTL mouse monoclonal antibody has been tested in western blot against recombinant transfected protein and in paraffin embedded immunohistochemistry on human spleen tissue sections.

Immunogen

Purified recombinant fragment of human ARNTL expressed in E. Coli.

Application

Anti-ARNTL Antibody, clone 1C5 is a highly specific mouse monoclonal antibody, that targets ARNT-Like 1 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected ARNTL in human spleen tissue.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in ARNTL-hIgGFc transfected HEK293 cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected ARNTL-hIgGFc transfected HEK293 cell lysate.

Target description

~100 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
ARNTL-hIgGFc transfected HEK293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service