Skip to Content
Merck
All Photos(1)

Key Documents

MABN642

Sigma-Aldrich

Anti-GST Omega 1 (GSTO1), clone 1H10.1 Antibody

clone 1H10.1, from mouse

Synonym(s):

Glutathione S-transferase omega-1, GSTO-1, Glutathione S-transferase omega 1-1, GSTO 1-1, Glutathione-dependent dehydroascorbate reductase, Monomethylarsonic acid reductase, MMA(V) reductase, S-(Phenacyl)glutathione reductase, SPG-R, GST Omega 1 (GSTO1)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$309.00

$309.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$309.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$309.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H10.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GSTO1(9446)

General description

GST Omega 1 (GSTO1), also known as Glutathione S-transferase omega-1,GSTO-1, Glutathione S-transferase omega 1-1, GSTO 1-1, Glutathione-dependent dehydroascorbate reductase, Monomethylarsonic acid reductase, MMA(V), or S-(Phenacyl)glutathione reductase, SPG-R, and encoded by the gene GSTO1/GSTTLP28, a ubiquitously expressed cytoplasmic enzyme that is highest in the liver and lowest in the brain It belongs to the GST superfamily of anti-oxidant reducing enzymes and is in the omega class. The omega class of glutathione transferases (GSTs) is a relatively ancient member of the cytosolic GST superfamily, and the omega-class GSTs are found in plants, animals, and some microbial species. The omega-class, unlike other GSTs, has a cysteine residue in their active site. Consequently, the omega-class GSTs catalyze a range of thiol transferase and reduction reactions that are not catalyzed by members of the other classes. GST Omega 1 (GSTO1) is involved in L-ascorbic acid metabolism, detoxification of arsenic substances, glutathione biosynthetic processes, regulation of ryanodine-sensitive calcium release channels, regulation of skeletal muscle contraction and the release of sequestered calcium in muscle, and xenobiotic catabolism. Interestingly genetic linkage studies have discovered associations between GSTO1 gene mutations and the age at onset of Alzheimer′s disease, Parkinson′s disease, and amyotrophic lateral sclerosis, perhaps stemming from the capacity of omega-class GSTs to mitigate oxidative stress or their role in activating the proinflammatory cytokine, interleukin-1β.

Immunogen

GST-tagged recombinant protein corresponding to human GST Omega 1 (GSTO1).

Application

Immunohistochemistry Analysis: A 1:50-500 dilution from a representative lot detected GST Omega 1 (GSTO1) in human cerebral cortex and rat cerebellum tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-GST Omega 1 (GSTO1), clone 1H10.1 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of GST Omega 1 (GSTO1).

Quality

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected GST Omega 1 (GSTO1) in 200 µg of human heart tissue lysate.

Target description

~28 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geeng-Fu Jang et al.
Cancers, 13(14) (2021-07-25)
Uveal melanoma metastases are lethal and remain incurable. A quantitative proteomic analysis of 53 metastasizing and 47 non-metastasizing primary uveal melanoma (pUM) was pursued for insights into UM metastasis and protein biomarkers. The metastatic status of the pUM specimens was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service