Skip to Content
Merck
All Photos(3)

Key Documents

MABN630

Sigma-Aldrich

Anti-SDHA Antibody, clone 7H11.2

clone 7H11.2, from mouse

Synonym(s):

Flavoprotein subunit of complex II, Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$599.00

$599.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$599.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$599.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7H11.2, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SDHA(6389)

General description

SDHA, also known as Succinate dehydrogenase flavoprotein-mitochondrial subunit, or Flavoprotein subunit of complex II (Fp), and encoded by the gene symbol SDHA/ SDH2/ SDHF, is a member of the FAD-dependent oxidoreductase 2 family and the FRD/SDH subfamily. Flavoprotein (FP) subunit of succinate dehydrogenase (SDH) is involved in the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q). Recent studies have indicated that SDHA may act as a tumor suppressor. SDHA is composed of four subunits: the flavoprotein (FP) SDHA, iron-sulfur protein (IP) SDHB, and a cytochrome b560 composed of SDHC and SDHD. SDHA interacts with SDHAF2/SDH5 in the process required for FAD attachment. Additionally, FDHA has been shown to interact with TRAP1. Mutations in the SDHA gene have been associated with Mitochondrial complex II deficiency (MT-C2D), a disorder of the mitochondrial respiratory chain with heterogeneous clinical manifestations. Additionally, mutations in the SDHA gene have been implied in Leigh syndrome or Kearns-Sayre syndrome, Cardiomyopathy dilated 1GG (CMD1GG), and Paragangliomas 5 (PGL5).

Immunogen

Epitope: Immunogen is near the C-terminal.
GST-tagged recombinant protein corresponding to the C-terminus of Human SDHA.

Application

Anti-SDHA, clone 7H11.2 is an antibody against SDHA for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blotting Analysis: A 0.5 µg/mL dilution from a representative lot detected SDHA in 10 µg of C2C12 and HeLa cell lysates.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected SDHA in human cerebral cortex and human cardiac myocytes tissues.

Quality

Evaluated by Western Blotting in Human brain tissue lysate.

Western Blotting Analysis: A 0.5 µg/mL diultion of this antibody detected SDHA in 10 µg of Human brain tissue lysate.

Target description

~68 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service