Skip to Content
Merck
All Photos(1)

Key Documents

MABN629

Sigma-Aldrich

Anti-PPID Antibody, clone 6A5.1

clone 6A5.1, from mouse

Synonym(s):

Peptidyl-prolyl cis-trans isomerase D, PPIase D, 40 kDa peptidyl-prolyl cis-trans isomerase, Cyclophilin-40, CYP-40, Cyclophilin-related protein, Rotamase D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$699.00

$699.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$699.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$699.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6A5.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPID(5481)

General description

PPIase D is one of a family peptidyl-prolyl-isomerases that help proteins fold during synthesis. Specifically, PPIases D catalyzes the cis-trans isomerization of proline imidic peptide bonds in peptides. Among the proteins assisted by PPIase D are HSP90, Estrogen receptor, cellular dynein, MYB, ILF2, XRCC6, GNB2L1 and RPS3. PPIase D also may regulate the interactions of other proteins including MYB and AHR. Through its protein folding exercise PPIase D becomes vital for proper protein transport and expression as well. Finally PPIase D is associated with assisting in the response to UV light induced apoptosis, cell viability in anaplastic lymphoma and hepatitis C virus replication and release. PPIase can be found localized in both the cytoplasm and the nucleus and is widely expressed. Note PPIase should not be confused with PPIF, or mitochondrial peptidyl-prolyl cis-trans isomerase F.

Immunogen

Recombinant protein corresponding to human PPID.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PPID in rat cerebellum tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-PPID Antibody, clone 6A5.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of PPID.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected PPID in 10 µg of HepG2 cell lysate.

Target description

~ 42 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service