Skip to Content
Merck
All Photos(2)

Key Documents

MABN616

Sigma-Aldrich

Anti-Potassium Channel Kv1.1 Antibody, clone K36/15

clone K36/15, from mouse

Synonym(s):

Potassium voltage-gated channel subfamily A member 1, RBKI, RCK1, Voltage-gated potassium channel subunit Kv1.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

K36/15, monoclonal

species reactivity

mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Potassium voltage-gated channel subfamily A member 1, better known as Potassium Channel Kv1.1, or Kcna1 belongs to the Shaker potassium channel group responsible for mediating ion permeability of excitable membranes. Conformational changes as a result of voltage changes across the cell membrane varies the permeability of this multi-pass transmembrane channel to potassium. Mutations of the Shaker potassium channel gene in Drosophila and the KCNA1 gene for its mammalian orthologue, Kv1.1, reduce the probability of action potential failure thereby causing hyperexcitability near axon branch points and nerve terminals, thereby leading to uncontrolled movements and recapitulating the episodic ataxia-1 (EA1) symptoms in human patients. Further, the axonal Kv1 channel activity may regulate synaptic transmission by modulating the action potential waveform, duration, firing pattern, and spread of depolarization into axonal branches.

Specificity

Other homologies: Human (94% sequence homology).

Immunogen

Linear peptide corresponding to 18 amino acids from the extracellular domain of rat Potassium Channel Kv1.1.

Application

Anti-Potassium Channel Kv1.1 Antibody, clone K36/15 is a highly specific mouse monoclonal antibody, that targets Kv1.1 & has been tested in Western blotting, Immunofluorescence & IHC.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western Blotting Analysis: 4 µg/mL from a representative lot detected Potassium Channel Kv1.1 in 10 µg of rat brain membrane tissue lysate.

Immunofluoresense Analysis: A representative lot from an independent laboratory detected Potassium Channel Kv1.1 in Kv1.1 transfected COS-1 cells (Tiffany, A. M., et al. (2000). J Cell Biol. 148(1):147-158.).

Immunhistochemistry Analysis: A representative lot from an independent laboratory detected Potassium Channel Kv1.1 in Kv1.1 transfected COS-1 cells (Wenzel. H. J., et al (2007). Epilepsia. 48(11):2023-2046.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detect Potassium Channel Kv1.1 in wild type mouse hippocampus tissue, and demonstrated a loss of signal in Potassium Channel Kv1.2 knock out mouse hippocampus tissue (Prof. J. Trimmer, University of California, Davis.).

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Potassium Channel Kv1.1 in 10 µg of mouse brain tissue lysate.

Target description

~65 kDa observed. The calculated molecular weight of this protein is 56 kDa However, it can run higher due to glycosylation (Deal, K. K., et al. (1994). J Neurosci. 14(3 Pt 2):1666-1676.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service