Skip to Content
Merck
All Photos(3)

Key Documents

MABN527

Sigma-Aldrich

Anti-iNOS/NOS II, clone 13F5.1 Antibody

culture supernatant, clone 13F5.1, from mouse

Synonym(s):

Nitric oxide synthase, inducible, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, iNOS, NOS type II, Peptidyl-cysteine S-nitrosylase NOS2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$803.00

$803.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$803.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$803.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

13F5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1λ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NOS2(4843)

General description

iNOS/NOS II, also known as Nitric oxide synthase inducible, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, iNOS, NOS type II, Peptidyl-cysteine S-nitrosylase , and encoded by the gene name NOS2 and NOS2A, produces nitric oxide (NO) which is a messenger molecule with diverse functions throughout the body. Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) & endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. Cytokines such as interferon-gamma (IFN), tumor necrosis factor (TNF), interleukin-1 and -2, and lipopolysaccarides (LPS) cause an increase in iNOS mRNA, protein, and activity levels. Protein kinase C-stimulating agents exhibit the same effect on iNOS activity. Human iNOS is regulated by calcium/calmodulin (in contrast with mouse NOS2). iNOS/NOS II is expressed in the liver, retina, bone cells and airway epithelial cells of the lung.

Immunogen

GST-tagged recombinant protein corresponding to human iNOS/NOS II.

Application

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected iNOS/NOS II in human cerebral cortex, human cerebellum, and human colon tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-iNOS/NOS II antibody is validated for use in WB, IH for the detection of iNOS/NOS II.

Quality

Evaluated by Western Blotting in human small intestine tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected iNOS/NOS II in 10 µg of human small Intestine tissue lysate.

Target description

~130 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Mouse monoclonal IgG1λ in buffer containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang Mi Park et al.
International journal of molecular medicine, 41(5), 2771-2783 (2018-02-14)
Socheongryong-Tang (SCRT) is a natural medicine prescription that has been mainly used in East Asia for the treatment of inflammatory disorders, including asthma and allergic rhinitis. The present study evaluated the anti-inflammatory effects of SCRT on lipopolysaccharide (LPS)-stimulated RAW 264.7 cells
Sonja Stojković Burić et al.
Oxidative medicine and cellular longevity, 2019, 3061607-3061607 (2019-04-16)
The main reasons for the inefficiency of standard glioblastoma (GBM) therapy are the occurrence of chemoresistance and the invasion of GBM cells into surrounding brain tissues. New therapeutic approaches obstructing these processes may provide substantial survival improvements. The purpose of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service