Skip to Content
Merck
All Photos(2)

Key Documents

MABN476

Sigma-Aldrich

Anti-Paris/ZNF746 Antibody, clone N196/16

clone N196/16, from mouse

Synonym(s):

Zinc finger protein 746, Parkin-interacting substrate, PARIS

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$616.00

$616.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$616.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$616.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N196/16, monoclonal

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZNF746(155061)

General description

Zinc finger protein 746 (ZNF746) is also called Parkin-interacting substrate (PARIS). ZNF746 is a repressor of transcription that binds to promoters and represses the transcription of PGC-1-alpha (PPARGC1A), making it important to the regulation of neuron death. ZNF746 may also contribute to neurodegeneration in Parkinson disease.

Specificity

This antibody recognizes the N-terminus of ZNF746. This antibody does not cross react with ZNF783 or ZNF398 (Prof. J. Trimmer, University of California, Davis.).

Immunogen

Epitope: N-terminus
Recombinant protein corresponding to the N-terminus of human Paris/ZNF746.

Application

Anti-Paris/ZNF746 Antibody, clone N196/16 is a highly specific mouse monoclonal antibody & that targets Zinc Finger Protein & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blotting Analysis: 1 µg/mL of this antibody detected Paris/ZNF746 in 10 µg of RAW264.7 cell lysate. Immunohistochemistry Analysis: A representative lot detected Paris/ZNF746 in rat brain tissue (Prof. J. Trimmer, University of California, Davis.).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Paris/ZNF746 in 10 µg of HEK293 cell lysate.

Target description

~72 kDa observed. Uniprot describes 3 isoforms, two at 69 kDa and one at 49 kDa This protein is ubiquitinated and has been observed at higher molecular weights (Shin, J. H., et al. (2011). Cell. 144(5):689-702.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi-Ruei Huang et al.
Journal of cellular biochemistry, 125(2), e30507-e30507 (2023-12-04)
The underlying mechanism of colorectal cells developing into cancer cells has been extensively investigated, yet is still not fully delineated, resulting in the treatment of advanced colorectal cancer (CRC) remains regrettably an unmet need. Zinc Finger Protein 746/Parkin-interacting substrate (ZNF746/PARIS)
Benjamin P Woodall et al.
JCI insight, 5 (2019-04-17)
The E3 ubiquitin ligase Parkin plays an important role in regulating clearance of dysfunctional or unwanted mitochondria in tissues, including the heart. However, whether Parkin also functions to prevent cardiac aging by maintaining a healthy population of mitochondria is still
Ju-Hyeon Bae et al.
Cell death & disease, 11(5), 359-359 (2020-05-14)
Various stresses, including oxidative stress, impair the proliferative capacity of muscle stem cells leading to declined muscle regeneration related to aging or muscle diseases. ZNF746 (PARIS) is originally identified as a substrate of E3 ligase Parkin and its accumulation is
Milena Pinto et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(4), 1042-1053 (2017-12-10)
PARK2 is the most common gene mutated in monogenic recessive familial cases of Parkinson's disease (PD). Pathogenic mutations cause a loss of function of the encoded protein Parkin. ParkinKO mice, however, poorly represent human PD symptoms as they only exhibit
Hojin Kang et al.
Oncotarget, 8(30), 48603-48618 (2017-06-15)
Progressive dopaminergic neurodegeneration is responsible for the canonical motor deficits in Parkinson's disease (PD). The widely prescribed anti-diabetic medicine metformin is effective in preventing neurodegeneration in animal models; however, despite the significant potential of metformin for treating PD, the therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service