Skip to Content
Merck
All Photos(1)

Key Documents

MABN445

Sigma-Aldrich

Anti-Synaptotagmin-like protein 1 (SYTL1 Antibody), clone 2F2.2

clone 2F2.2, 1 mg/mL, from mouse

Synonym(s):

Synaptotagmin-like protein 1, Exophilin-7, Protein JFC1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$524.00

$524.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$524.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$524.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F2.2, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYTL1(84958)

General description

Synaptotagmin-like protein 1 (SYTL1), also known as Exophilin-7, Protein JFC1, and encoded by the gene SYTL1/SLP1, is an important protein in exocytosis. Synaptotagmin-like protein 1 plays a role in vesicle trafficking and is important in exocytosis in lymphocytes and other cells including neurons, platelets and most cells that secrete materials via exocytosis. Synaptotagmin-like protein 1 is highly expressed in bone marrow and lymphoid tissues as well as cerebellum, occipital lobe, prostate, stomach, kidney, appendix, lung and trachea.

Immunogen

GST-tagged recombinant protein corresponding to human Synaptotagmin-like protein 1 (SYTL1).

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Synaptotagmin-like protein 1 (SYTL1) in human cerebral cortex and human tonsil tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Synaptotagmin-like protein 1 (SYTL1 Antibody), clone 2F2.2 is validated for use in western blotting & IHC for the detection of Synaptotagmin-like protein 1 (SYTL1).

Quality

Evaluated by Western Blotting in human lymph node tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Synaptotagmin-like protein 1 (SYTL1) in 10 µg of human lymph node tissue lysate.

Target description

~62 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service