Skip to Content
Merck
All Photos(3)

Key Documents

MABN372

Sigma-Aldrich

Anti-CLN3 Antibody, Clone 2B8.2

clone 2B8.2, from mouse

Synonym(s):

Battenin, Batten disease protein, Protein CLN3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$699.00

$699.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$699.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$699.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B8.2, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLN3(1201)

General description

CLN3 protein is involved in lysosomal function. Mutations in this, as well as other neuronal ceroid-lipofuscinosis (CLN) genes, cause neurodegenerative diseases commonly known as Batten disease or collectively known as neuronal ceroid lipofuscinoses (NCLs).

Immunogen

KLH-conjugated linear peptide corresponding to human CLN3.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-CLN3 Antibody, Clone 2B8.2 is validated for use in Western Blotting and Immunohistochemistry for the detection of CLN3.
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected CLN3 in 10 µg of A431 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CLN3 in rat cerebellum and human thalamus tissue.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected CLN3 in 10 µg of HepG2 cell lysate.

Target description

~45 kDa observed. Uniprot describes 5 isoforms produced by alternative splicing at ~45, ~42, ~37, ~35, and ~28 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service