Skip to Content
Merck
All Photos(2)

Key Documents

MABN355

Sigma-Aldrich

Anti-HAND2 Antibody, Clone 7D4.1

clone 7D4.1, from mouse

Synonym(s):

Heart- and neural crest derivatives-expressed protein 2, Class A basic helix-loop-helix protein 26, bHLHa26, Deciduum, heart, autonomic nervous system and neural crest derivatives-expressed protein 2, dHAND

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$599.00

$599.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$599.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$599.00


Estimated to ship on07 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D4.1, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HAND2(9464)

General description

Heart-and neural crest derivatives-expressed protein 2 , also known as Class A basic helix-loop-helix protein 26 (bHLHa26), or Deciduum, heart, autonomic nervous system and neural crest derivatives-expressed protein 2 (dHAND), and encoded by the gene HAND2/BHLHA26/DHAND, is a transcription factor that is essential for cardiac morphogenesis, particularly the formation of the right ventricle and the aortic arch arteries. Heart-and neural crest derivatives-expressed protein 2 is required for vascular development, angiogenesis and for limb development acting upstream of sonic hedgehog in the induction of the limb bud. Additionally, Heart-and neural crest derivatives-expressed protein 2 is central to cardiac neural crest cell development and involved in outflow tract morphogenesis as well as the maintenance of the adult heart and in the negative regulation of cardiac muscle cell apoptosis. Heart-and neural crest derivatives-expressed protein 2 is localized to the nucleus. Heart-and neural crest derivatives-expressed protein 2 is expressed in developing heart, lung and vascular system.

Immunogen

Epitope: C-terminus outside the bHLH domain
Recombinant protein corresponding to the C-terminus outside the bHLH domain of human HAND2.

Application

Detect HAND2 using this Anti-HAND2 Antibody, Clone 7D4.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected HAND2 in human placenta tissue.

Quality

Evaluated by Western Blotting in mouse heart tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected HAND2 in 10 µg of mouse heart tissue lysate.

Target description

~30 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service