Skip to Content
Merck
All Photos(2)

Key Documents

MABN287

Sigma-Aldrich

Anti-ABCA4 Antibody, clone 5D7.1

clone 5D7.1, from mouse

Synonym(s):

Retinal-specific ATP-binding cassette transporter, ATP-binding cassette sub-family A member 4, RIM ABC transporter, RIM protein, RmP, Stargardt disease protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$589.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5D7.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCA4(24)

General description

Retinal-specific ATP-binding cassette transporter encoded by the gene ABCA4 plays a critical role in the transport of retinoid substrates from the extracellular or intradiscal rod cell membranes to the cytoplasmic membrane surface for processing ultimately into vitamin A and then transferred to the retinal pigment epithelium where it is converted to 11-cis-retinal that active component needed for vision. ABCA4 is localized to the outer segments of rods and cones with a million copies per photoreceptor. ABCA4 is retinal specific protein. Mutations in ABCA4 are linked to several eye-related genetic diseases, such as forms of macular degeneration called Stargardt disease and ARMD2, as well as Retinitis pigmentosa 19, and Cone-rod dystrophy 3.

Immunogen

Recombinant protein corresponding to human ABCA4.

Application

Anti-ABCA4 Antibody, clone 5D7.1 | MABN287 is an antibody against ABCA4 Antibody for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected ABCA4 in human retina tissue.

Quality

Evaluated by Western Blotting in Y79 cell lysate.

Western Blotting Analysis: 5.0 µg/mL of this antibody detected ABCA4 in 200 µg of Y79 cell lysate.

Target description

~275 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service