Skip to Content
Merck
All Photos(1)

Key Documents

MABN2617

Sigma-Aldrich

Anti-CNGA1/CNG1 Antibody, clone PMc 1D1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$300.00
100 μG
$753.00

$300.00


Estimated to ship on28 May 2025



Select a Size

Change View
25 μG
$300.00
100 μG
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$300.00


Estimated to ship on28 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies, primary antibodies

clone

PMc 1D1, monoclonal

mol wt

calculated mol wt 79.6 kDa
observed mol wt ~79.6 kDa

purified by

using protein G

species reactivity

bovine, mouse, rat

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

bovine ... CNGA1(281700)

General description

cGMP-gated cation channel alpha-1 (UniProt: Q00194; also known as Cyclic nucleotide-gated cation channel 1, Cyclic nucleotide-gated channel alpha-1, CNG channel alpha-1, CNG-1, CNG1, Cyclic nucleotide-gated channel, photoreceptor, Rod photoreceptor cGMP-gated channel subunit alpha) is encoded by the CNGA1 (also known as CNCG, CNCG1) gene (Gene ID: 281700) in bovine species. CNGA1 is a multi-pass membrane protein with four cytoplasmic domains, six transmembrane domains, and three extracellular domains. It is a subunit of the rod cyclic GMP-gated cation channel that is localized to the rod outer segment and is involved in the final stage of the phototransduction pathway. It forms a heterotetramer with CNGB1 (in a 3;1 ratio) but may also form homotetrameric channels that are strongly activated by saturating cGMP. When light hits rod photoreceptors, cGMP levels decrease that results in rapid closure of CNGA1/CNGB1 channels and hyperpolarization of the membrane potential. In this channel, CNGA1 forms a central channel gate in the middle of the membrane and occludes the central cavity. CNGA1 has two structural components: the membrane-spanning S1-S6 region and the cytosolic ligand gating apparatus. The transmembrane S1-S6 region of each subunit is separated into an S1-S4 voltage-sensor domain and an S5-S6 pore domain. The cytosolic soluble fragment of CNGA1 forms the gating apparatus in a channel tetramer that is responsible for cGMP activation of the channel. (Ref.: Xue, J., et al. (2021). Neuron. 109(8); 1302-1313; Cook, NJ., et al. (1989). J. Biol. Chem. 264(12); 6996-6999).

Specificity

Anti-CNGA1/CNG1, clone PMc 1D1, Cat. No. MABN2617, is a mouse monoclonal antibody that detects cGMP-gated cation channel alpha-1 (CNGA1) and is tested for use in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Immunogen

Plasma membrane preparation purified from bovine rod outer segments.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in mouse retina tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected CNGA1/CNG1 in mouse retina tissue sections.

Tested Applications

Immunohistochemistry Applications: A representative lot detected CNGA1/CNG1 in Immunohistochemistry applications (Sharif, A.S., et al. (2018). J Neurosci. 38(13):3160-3176).

Immunoprecipitation Analysis: A representative lot detected CNGA1/CNG1 in Immunoprecipitation applications (Cheng, C.L., et al. (2013). J Cell Sci. 126(Pt24):5725-34).

Western Blotting Analysis: A representative lot detected CNGA1/CNG1 in Western Blotting applications (Cook, N.J., et al. (1989). J Biol Chem. 264(1):6996-9; Cheng, C.L., et al. (2013). J Cell Sci. 126(Pt24):5725-34).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
This mouse monoclonal Anti-CNGA1/CNG1, clone PMc 1D1, Cat. No. MABN2617 is tested for use in Immunohistochemistry (Paraffin), Western Blotting, Immunoprecipitation, for the detection of CNGA1.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service