Skip to Content
Merck
All Photos(1)

Key Documents

MABN2548

Sigma-Aldrich

Anti-CLN6 Antibody, clone N400/24

clone N400/24, from mouse

Synonym(s):

Ceroid-lipofuscinosis neuronal protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$252.00
100 μG
$735.00

$252.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$252.00
100 μG
$735.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N400/24, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CLN6(54982)

General description

Ceroid-lipofuscinosis neuronal protein 6 (UniProt: Q9NWW5; also known as CLN6) is encoded by the CLN6 gene (Gene ID: 54982) in human. CLN6 is a multi-pass membrane protein that is localized to endoplasmic reticulum and contributes to lysosomal function and viability of neurons. It has no homology with known proteins or functional domains, however it is highly conserved across mammalian species. Mutations in CLN6 gene have been linked to ceroid lipofuscinosis, neuronal 6 and 4A that are characterized by intracellular accumulation of autofluorescent liposomal material, seizures, dementia, visual loss, and/or cerebral atrophy. Two isoforms of CLN6 have been described that are produced by alternative splicing.

Specificity

Clone N400/24 specifically detects human Ceroid-lipofuscinosis neuronal protein 6. It does not cross-react with other CLN proteins.

Immunogen

Recombinant fragments corresponding to 54, 47, and 30 amino acids from non-helical regions from the N-terminal, internal, and C-terminal regions of human Ceroid-lipofuscinosis neuronal protein 6 (CLN6).

Application

Anti-CLN6, clone N400/24, Cat. No. MABN2548, is a highly specific mouse monoclonal antibody that targets CLN6 and has been tested for use in Immunohistochemistry (Paraffin).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry (Paraffin) in human stomach tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected CLN6 in human stomach tissue sections.

Target description

35.92 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service