Skip to Content
Merck
All Photos(1)

Key Documents

MABN2429M

Sigma-Aldrich

Anti-GARP Antibody, clone 4B1

clone 4B1, from mouse

Synonym(s):

Cyclic nucleotide-gated cation channel beta-1, Cyclic nucleotide-gated cation channel 4, CNG channel 4, CNG-4, CNG4, Cyclic nucleotide-gated cation channel gamma, Cyclic nucleotide-gated cation channel modulatory subunit, Cyclic nucleotide-gated channel

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00
100 μG
$520.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$214.00
100 μG
$520.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B1, monoclonal

species reactivity

bovine, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), human (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... LRRC32(2615)

General description

Cyclic nucleotide-gated cation channel beta-1 (UniProt: Q14028; also known as Cyclic nucleotide-gated cation channel 4, CNG channel 4, CNG-4, CNG4, Cyclic nucleotide-gated cation channel gamma, Cyclic nucleotide-gated cation channel modulatory subunit, Cyclic nucleotide-gated channel beta-1, CNG channel beta-1, CNGB1, Glutamic acid-rich protein, GARP) is encoded by the CNGBI (also known as CNCG2, CNCG3L, CNCG4, RCNC2) gene in human. CNGB1 is a multi-pass membrane protein that is a subunit of cyclic nucleotide-gated (CNG) channels that are non-selective cation channels and play important roles in both visual and olfactory signal transduction. It forms a tetramer with three CNGA1 subunits. When associated with CNGA1, it is involved in the regulation of ion flow into the rod photoreceptor outer segment (ROS), in response to light-induced alteration of the levels of intracellular cGMP. CNGB1 contains one cyclic nucleotide-binding domain. Four isoforms of CNGB1 have been described that are generated by alternate splicing. Mutations in CNGB1 gene are associated with retinitis pigmentosa that is characterized by retinal pigment deposits that are visible on fundus examination and case loss of rod photoreceptor cells followed by secondary loss of cone photoreceptors.

Specificity

Clone 4B1 detects Glutamic acid-rich protein (GARP) in mouse and bovine retina. It targets an epitope with in the first 16 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
GST-tagged first 16 amino acids from the N-terminal region of human Cyclic nucleotide-gated cation channel beta-1.

Application

Anti-GARP, clone 4B1, Cat. No. MABN2429, is a highly specific mouse monoclonal antibody, that targets CNGB1 and has been tested in Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected GARP in Western Blotting applications (Poetsch, A., et. al. (2001). J Biol Chem. 276(51):48009-16; Chakraborty, D., et. al. (2008). Biochemistry. 47(4):1144-56; Zulliger, R., et. al. (2015). PLoS One. 10(9):e0138508).

Immunofluorescence Analysis: A representative lot detected GARP in Immunofluorescence applications (Zhang, Y., et. al. (2009). J Cell Sci. 122(Pt 8):1192-200).

Immunoprecipitation Analysis: A representative lot detected GARP in Immunoprecipitation applications (Conley, S.M., et. al. (2010). Adv Exp Med Biol. 664:63-70).

Quality

Evaluated by Immunohistochemistry in mouse retina tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected GARP in mouse retinal tissue.

Target description

139.68 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service