Skip to Content
Merck
All Photos(3)

Key Documents

MABN2397

Sigma-Aldrich

Anti-MYCBP2 Antibody, clone PM-8A9

clone PM-8A9, from mouse

Synonym(s):

E3 ubiquitin-protein ligase MYCBP2, Myc-binding protein 2, Pam/highwire/rpm-1 protein, Protein associated with Myc

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$574.00

$574.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$574.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$574.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PM-8A9, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MYCBP2(23077)

General description

E3 ubiquitin-protein ligase MYCBP2 (UniProt: O75592; also known as Myc-binding protein 2, Pam/highwire/rpm-1 protein, Protein associated with Myc, MYCBP2) is encoded by the MYCBP2 (also known as KIAA0916, PAM) gene (Gene ID: 23077) in human.MYCBP2 protein mediates the ubiquitination and subsequent proteasomal degradation of TSC2/tuberin. It is ubiquitously expressed, but brain and thymus display exceptionally abundant expression. It is shown to co-localize with TSC1 and TSC2 along the neurites and in the growth cones. MYCBP2 may also function as a facilitator or regulator of transcriptional activation by MYC and have a role during synaptogenesis. It is reported to negatively regulate neuronal growth, synaptogenesis, and synaptic strength. Mice with MYCBP2-deficiency in peripheral sensory neurons show prolonged thermal hyperalgesia. Loss of MYCBP2 is also linked to constitutively activated p38 MAPK and increased expression of several proteins involved in receptor trafficking. Down-regulation of MYCBP2 in the spinal cord of adult rats is shown to enhance their. MYCBP2 is also shown to be an inhibitor of adenylyl cyclase. (Ref.: Holland, S et al. (2011). J. Biol. Chem. 286(5): 3671-3680).

Specificity

Clone PM-8A9 specifically detects MYCBP2 in HEK293 cells. It targets an epitope within the first 150 amino acids in the N-terminal region.

Immunogen

Epitope: N-terminus
MBP (Maltose-binding protein)-tagged recombinat fragment of 150 amino acids from the N-terminal region of human MYCBP2.

Application

Anti-MYCBP2 Antibody, clone PM-8A9, Cat. No. MABN2397, is a highly specific mouse monoclonal antibody that targets MYCBP2 (PAM) and has been tested in Immunoprecipitation and Western blotting.
Immunoprecipitation Analysis: A representative lot detected MYCBP2 (Courtesy of Roberta L. Beauchamp, Massachusetts General Hospital, Boston, USA).

Western Blotting Analysis: A representative lot detected endogenous PAM (MYCBP2)and overexpressed Myc-tagged PAM (Courtesy of Roberta L. Beauchamp, Massachusetts General Hospital, Boston, USA).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HEK293 cell lysates.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MYCBP2 in 10 µg of HEK293 cell lysate.

Target description

~460 kDa observed; 510.08 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Fang et al.
Signal transduction and targeted therapy, 9(1), 218-218 (2024-08-23)
Obesity is a global issue that warrants the identification of more effective therapeutic targets and a better understanding of the pivotal molecular pathogenesis. Annexin A1 (ANXA1) is known to inhibit phospholipase A2, exhibiting anti-inflammatory activity. However, the specific effects of
Gongsheng Yuan et al.
International journal of oral science, 15(1), 11-11 (2023-02-17)
Tumor-associated macrophages (TAMs) play crucial roles in tumor progression and immune responses. However, mechanisms of driving TAMs to antitumor function remain unknown. Here, transcriptome profiling analysis of human oral cancer tissues indicated that regulator of G protein signaling 12 (RGS12)
Gongsheng Yuan et al.
Molecular therapy. Nucleic acids, 31, 197-210 (2023-01-27)
Synovial fibroblasts are the active and aggressive drivers in the progression of arthritis, but the cellular and molecular mechanisms remain unknown. Here, our results showed that regulator of G protein signaling 12 (RGS12) maintained ciliogenesis in synovial fibroblasts, which is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service