Skip to Content
Merck
All Photos(4)

Key Documents

MABN2286

Sigma-Aldrich

Anti-Rai Antibody, clone IF15

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$310.00
100 μL
$780.00

$310.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$310.00
100 μL
$780.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$310.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

IF15-E2, monoclonal

mol wt

calculated mol wt 64.06 kDa
observed mol wt ~58 kDa

species reactivity

human, mouse

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

SHC-transforming protein 3 (UniProt: Q92529; also known as Neuronal Shc, N-Shc, Protein Rai, SHC-transforming protein C, Src homology 2 domain-containing-transforming protein C3, SH2 domain protein C3, Rai) is encoded by the SHC3 (also known as NSHC, SHCC) gene (Gene ID:53358) in human. Rai protein is mainly expressed in the brain and its higher expression is observed in cerebral cortex, frontal and temporal lobes, hippocampus, and occipital pole. Its expression is also reported in the enteric glial cells in the human gut. It serves as a signaling adapter and is involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons. Rai also serves as a physiological substrate of the receptor tyrosine kinase Ret and increases Ret-dependent survival via PI3 kinase/Akt pathway activation. It also potentiates MAP kinase signaling. Rai activation also plays a role in long-term potential and is neuroprotective against certain ischemia and oxidative stress. Evolutionarily it correlates well with vertebrate brain development. Its reduced levels are observed in brain of Alzheimer s disease subjects. Two isoforms of Rai have been described (p64 and p52) that are produced by alternative splicing. (Ref.: Triaca, V., et al. (2018). Aging. 10(1); 5-6; Sagi, O., et al. (2015). Ageing Res. Rev. 19; 34-42; Villanacci, V., et al. (2008). Neurogastroenterol. Motil. 20(3); 206-212).

Specificity

Clone IF15-E2 is a mouse monoclonal antibody that detects SHC-transforming protein 3/RAI. It targets an epitope within the CH1 domain from the C-terminal half.

Immunogen

Recombinant fragment corresponding to 164 amino acids CH1 domain of human SHC-transforming protein 3 (Rai).

Application

Quality Control Testing

Evaluated by Western Blotting in Human brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Rai in Human brain tissue lysate.

Tested Applications

Immunohistochemistry Applications: A representative lot detected Rai in Immunohistochemistry applications (Villanacci, V., et. al. (2008). Neurogastroenterol Motil. 20(3):206-12).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Rai in U2OS cells.

Immunohistochemistry (Paraffin) Analysis: A 1:500 dilution from a representative lot detected Rai in human esophagus, stomach, and colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Rai, clone IF15, Cat. No. MABN2286, is a mouse monoclonal antibody that detects SHC-transforming protein 3 (Rai) and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Physical form

Purified mouse monoclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service