Skip to Content
Merck
All Photos(2)

Key Documents

MABN1793

Sigma-Aldrich

Anti-SorLA Antibody, clone 20C11

clone 20C11, from mouse

Synonym(s):

Sortilin-related receptor, Gp250, LDLR relative with 11 ligand-binding repeats, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11, SorLA, SorLA-1, Sorting protein-related receptor containing LDLR class A repeats

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$598.00

$598.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$598.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$598.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20C11, monoclonal

species reactivity

mouse, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SORL1(6653)

General description

Sortilin-related receptor (UniProt Q92673; also known as Gp250, LDLR relative with 11 ligand-binding repeats, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11, SorLA, SorLA-1, Sorting protein-related receptor containing LDLR class A repeats) is encoded by the SORL1 (also known as C11orf32) gene (Gene ID 6653) in human. SorLA and Sortilin are related Vps10p-domain (Vsp10p-D) family multi-ligand-binding receptors that target LPL, growth factors and peptides, as well as soluble amyloid precursor proein (sAPP) for endocytosis. Studies show that SorLA protects sAPP against proteolytic processing in compartments localized in the cell body (soma) of hippocampal neurons, whereas sortilin facilitates sAPP degradation by lysosomes localized in neurites. Hippocampal neurons lacking sortilin display reduced sAPPα production, suggesting that sortilin promotes α-secretase activity in neuronal systems. Unlike LRP1 and LRP1b, two other known sAPP-binding receptors, SorLA and Sortilin bind sAPP in a KPI domain-independent manner and their scavenging function therefore is not limited to sAPP produced from KPI-containing APP isoforms. Human SorLA is a single transmembrane (a.a. 2138-2158) protein with a 55-amino acid cytoplasmic tail (a.a. 2159-2214) and a large luminal/extracellular (a.a. 1-2137) region that contains a Vps10p-domain (Vsp10p-D; a.a. 1-731) with a signal peptide (a.a. 1-28) and a propeptide (a.a. 29-81) sequence at the N-terminal end and five BNR (bacterial neuraminidase repeat or (F/Y)WTD/b-propeller domain) repeats (a.a. 136-573). The C-terminal half of the luminal/extracellular region contains five LDL-R class B (LB) repeats (a.a. 800-1013), an EGF-like domain (a.a. 1026-1072), eleven LDL-R class A (LA) repeats (a.a. 1076-1551), and six fibronectin type-III repeats (a.a. 1557-2118).

Specificity

Clone 20C11 immunostained SorLA endocytic vesicles in wild-type, but not Sorl1-/-, mouse astrocytes.

Immunogen

Epitope: Extracellular/luminal region outside the Vps10p domain (Vsp10p-D).
Recombinant human SorLA extracellular/luminal domain.

Application

Anti-SorLA Antibody, clone 20C11 is an antibody against SorLA for use in Immunocytochemistry, Western Blotting, ELISA.
Immunocytochemistry Analysis: 5 µg/mL from a representative lot immunostained SorLA endocytic vesicles in wild-type, but not Sorl1-/-, mouse astrocytes (Courtesy of Dr. Jakob V. Larsen, MIND-centre, University of Aarhus, Denmark).
Immunocytochemistry Analysis: A representative lot detected SorLA immunoreactivity primarily located at the cell body (soma) by fluorescent immunocytochemistry staining of 4% paraformaldehyde-fixed, 0.1% Triton X-100-permeabilized primary murine hippocampal neurons (Gustafsen, C., et al. (2013). J. Neurosci. 33(1):64-71).
Immunocytochemistry Analysis: A representative lot immunostained endocytic vesicular structures containing endocytosed sAPP by fluorescent immunocytochemistry staining of 4% paraformaldehyde-fixed, 0.1% Triton X-100-permeabilized HEK293 cells expressing exogenously transfected human SorLA (Gustafsen, C., et al. (2013). J. Neurosci. 33(1):64-71).
Immunocytochemistry Analysis: A representative lot detected the cellular localization of exogenously expressed wild-type and the FANSHY-to-6A4 mutant human SorLA by fluorescent immuncytochemistry staining of 4% paraformaldehyde-fixed, 0.5% saponin-permeabilized SH-SY5Y transfectants (Fjorback, A.W., et al. (2012). J. Neurosci. 32(4):1467-1480).
Western Blotting Analysis: A representative lot detected endogenous SorLA in sucrose gradient-fractionated mouse brain extracts as well as the expression of exogenously transfected human SorLA in HEK293 transfectants (Gustafsen, C., et al. (2013). J. Neurosci. 33(1):64-71).
ELISA Analysis: Representative lots were employed as the detection antibody for quantiating SorLA level in human brain cortical extracts as well as the level of exogenously expressed human SorLA in CHO cells (Caglayan, S., et al. (2012). Arch. Neurol. 69(3):373-379; Schmidt, V., et al. (2012). EMBO J. 31(1):187-200).

Quality

Evaluated by Immunocytochemistry in human SorLA-expressing HEK293 cells.

Immunocytochemistry Analysis: 4.0 µg/mL of this antibody immunostained SorLA endocytic vesicles in HEK293 cells expressing transfected human SorLA.

Target description

239.3 kDa (human) and 238.3 kDa (mouse) calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillermo A Herrera et al.
Kidney international reports, 6(5), 1379-1396 (2021-05-21)
Deciphering the intricacies of the interactions of glomerulopathic Ig light chains with mesangial cells is key to delineate signaling events responsible for the mesangial pathologic alterations that ensue. Human mesangial cells, caveolin 1 (CAV1), wild type (WT) ,and knockout (KO)
Giulia Monti et al.
Journal of biotechnology, 375, 17-27 (2023-08-28)
Reduced levels of the Sortilin-related receptor with A-type repeats (SORLA) in different brain regions as well as in the cerebrospinal fluid have been associated with Alzheimer's disease. Methods and reagents to develop reliable detection assays to quantify SORLA and its
Paulina Kaminska et al.
EMBO reports, 25(5), 2278-2305 (2024-03-19)
SorLA, encoded by the gene SORL1, is an intracellular sorting receptor of the VPS10P domain receptor gene family. Although SorLA is best recognized for its ability to shuttle target proteins between intracellular compartments in neurons, recent data suggest that also
Peder Madsen et al.
Scientific reports, 9(1), 611-611 (2019-01-27)
SorLA and Sortilin are multifunctional receptors involved in endocytosis and intracellular sorting of different and unrelated ligands. SorLA has recently attracted much attention as a novel strong risk gene for Alzheimer's disease, and much effort is currently being put into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service