Skip to Content
Merck
All Photos(1)

Key Documents

MABN1597

Sigma-Aldrich

Anti-Olfactory Receptor 10G4 Antibody, clone 6C11.1

clone 6C11.1, from mouse

Synonym(s):

Olfactory receptor OR11-278, Olfactory Receptor 10G4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$422.00

$422.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$422.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$422.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C11.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Olfactory receptor 10G4 (UniProt Q8NGN3; also known as Olfactory receptor OR11-278) is encoded by the OR10G4 gene (Gene ID 390264) in human. Olfactory receptors belong to the class A rhodopsin-like family of G protein-coupled receptors (GPCRs). The human genome contains approximately 800 odorant receptor (OR) genes that have been shown to exhibit high genetic variability. Human has approximately 400 intact ORs, but each individual has a unique set of genetic variations that lead to variation in olfactory perception. Odorant molecule binding induces an OR conformational change and interaction with the olfactory-type G protein, initiating downstream signaling that eventually results in an action potential from the depolarized OR neuron to the brain. Olfactory receptor 10G4 is a 311-amino acid protein with 4 extracellular domains (a.a. 1-23, 74-98, 160-196, 258-268), 7 transmembrane segments (a.a. 24-44, 53-73, 99-119, 139-159, 197-216, 237-257, 269-289), and 4 cytoplasmic regions (a.a. 45-52, 120-138, 217-236, 290-311).

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminal cytoplasmic tail of human Olfactory Receptor 10G4.

Application

Anti-Olfactory Receptor 10G4 Antibody, clone 6C11.1 is an antibody against Olfactory Receptor 10G4 for use in Western Blotting.

Quality

Evaluated by Western Blotting in human olfactory tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Olfactory Receptor 10G4 in 10 µg of human olfactory tissue lysate.

Target description

~40 kDa observed. Target band size appears larger than the calculated molecular weight of 34.57 kDa due to glycosylation.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service