Skip to Content
Merck
All Photos(1)

Key Documents

MABN1578

Sigma-Aldrich

Anti-GNAT1 Antibody, clone 11A8.1

clone 11A8.1, from mouse

Synonym(s):

Guanine nucleotide-binding protein G(t) subunit alpha-1, Transducin alpha-1 chain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$512.00

$512.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$512.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$512.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11A8.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... GNAT1(2779)

General description

Guanine nucleotide-binding protein G(t) subunit alpha-1 (UniProt P11488; also known as Transducin alpha-1 chain) is encoded by the GNAT1 (also known as GNATR, CSNB1G, CSNBAD3) gene (Gene ID 2779) in human. Heterotrimeric G proteins are activated by G protein-coupled receptors (GPCRs) and are composed of alpha (α), beta (β), and gamma (γ) subunits. There exists also monomeric Ras superfamily of small GTPases that are commonly referred to as small G-proteins. G-proteins mediate signal transduction initiated by GPCRs. Ligand binding activates GPCRs by inducing a conformational change. The activated GPCRs catalyze the GDP-to-GTP exchange of the Gα subunit, causing a dissociation of the Gβγ dimer from the now GTP-bound Gα subunit. The Gα subunit gradually hydrolyzes the bound GTP to GDP and eventually re-associates with Gβγ and re-starts the cycle. There exist at least 20 different Gα subunits, which are separated into four main groups, Gi, Gs, Gq, and G12/13. Eight different genes code for Gi alpha subunits, including GNAT1 and GNAT2 that code for Gt in the rod (rTα by GNAT1) and cone (cTα by GNAT2) cells of the retina, where they function as the alpha subunit of the trimeric G protein transducin. The α-subunit binds GTP and activates cyclic GMP phosphodiesterase (PDE), whereas the β and γ subunits mediate interaction with rhodopsin. Mutations in RHO, PDE6B, and GNAT1 genes are associated with autosomal dominant congenital stationary night blindness (CSNB), and patients with mutations in these genes exhibit the Riggs- or Schubert-Bornschein-type electronegative electroretinogram (ERG).

Specificity

Clone 11A8.1 was raised against human Gt alpha-1 subunit (GNAT1) recombinant fragment. Cross-reactivity toward human Gt alpha-2 subunit (GNAT2) has not been determined.

Immunogen

GST-tagged recombinant human GNAT1 internal fragment.

Application

Anti-GNAT1 Antibody, clone 11A8.1 is an antibody against GNAT1 for use in Western Blotting.

Quality

Evaluated by Western Blotting in human retina tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected GNAT1 in 10 µg of human retina tissue lysate.

Target description

~40 kDa observed. 39.91 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service