Skip to Content
Merck
All Photos(2)

Key Documents

MABN1537

Sigma-Aldrich

Anti-TDO2 Antibody, clone 7A6.1

clone 7A6.1, from mouse

Synonym(s):

Tryptophan 2, 3-dioxygenase, TDO, TO, TRPO, Tryptamin 2, 3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$564.00

$564.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A6.1, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TDO2(6999)

General description

Tryptophan 2,3-dioxygenase (EC 1.13.11.11; UniProt P48775; also known as TDO, TO, TRPO, Tryptamin 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase) is encoded by the TDO2 (also known as TDO) gene (Gene ID 6999) in human. TDO2 is a cytosolic heme-containing enzyme that exists as a homotetramer, where the N terminal residues of each monomer form part of the substrate binding site in an adjacent monomer. Tryptophan 2,3-dioxygenase (TDO2) catalyzes the first step of the kynurenine pathway, namely the conversion of tryptophan to kynurenine (Kyn), which can also be catalyzed by indoleamine 2,3-dioxygenase 1 (IDO1) or IDO2. IDO1 is expressed in tissues throughout the body, while TDO2 is predominantly expressed in the liver. Altered tryptophan metabolism and increased secretion of tryptophan metabolites are found in solid tumors, including in breast cancer. TDO2-mediated Kyn production in triple-negative breast cancer (TNBC) cells is reported to activate aryl hydrocarbon receptor (AhR), an endogenous kynurenine receptor. Inhibiting TDO2 or AhR via pharmacologic or genetic means is reported to result in increased sensitivity to anoikis, as well as decreased proliferation, migration, and invasion of TNBC cells.

Specificity

Clone 7A6.1 targets an internal epitope of human and murine TDO2 (TDO).

Immunogen

GST-tagged recombinant human TDO2 internal fragment.

Application

Anti-TDO2, clone 7A6.1, Cat. No. MABN1537, is a highly specific mouse monoclonal antibody that targets TDO2, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TDO2 in human kidney, liver, and small intestine tissue sections.

Quality

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected TDO2 in 10 µg of mouse liver tissue lysate.

Target description

~48 kDa observed. 47.87 kDa (human) and 47.76 kDa (mouse) calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeko Yoshioka et al.
International journal of tryptophan research : IJTR, 15, 11786469221138456-11786469221138456 (2022-12-06)
Kynurenine (Kyn), a metabolite of tryptophan (Trp), is a key regulator of mammal immune responses such as cancer immune tolerance. Indoleamine-2,3-dioxygenase (IDO) and tryptophan-2,3-dioxygenase (TDO) are main enzymes regulating the first and rate-limiting step of the Kyn pathway. To identify
Tatsuya Ando et al.
Molecular medicine reports, 27(2) (2022-12-10)
The partial loss of liver due to liver transplantation or acute liver failure induces rapid liver regeneration. Recently, we reported that the selective inhibition of indoleamine 2,3‑dioxygenase (Ido) 1 promotes early liver regeneration. However, the role of Ido2 in liver

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service