Skip to Content
Merck
All Photos(1)

Key Documents

MABF917

Sigma-Aldrich

Anti-CRACC Antibody, clone 4G2

clone 4G2, from rat

Synonym(s):

SLAM family member 7, CD2-like receptor-activating cytotoxic cells, CRACC, Leukocyte cell-surface antigen, Novel Ly9, CD319

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$598.00

$598.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$598.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

$598.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G2, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Slamf7 (75345)

Related Categories

General description

SLAM family member 7 (UniProt Q8BHK6; also known as CD2-like receptor-activating cytotoxic cells, CRACC, Leukocyte cell-surface antigen, Novel Ly9, CD319) is encoded by the Slamf7 (also known as Cs1) gene (Gene ID 75345) in murine species. CRACC/CS1 belongs to the 6-member (SLAM/CD150, Ly-9/CD229, 2B4/CD244, CD84, NTB-A/CD352, CRACC) signaling lymphocytic activation molecule family (SLAMF) of receptors broadly expressed in hematopoietic cells. 2B4 and CD48 interact with each other, while rest of the family members act as self-ligands, where two identical molecules on the surface of two different cells can interact as receptor and ligand to each other. CRACC is synthesized with a signal peptide sequence (a.a. 1-22), the removal of which yields the mature CRACC with an extracellular region (a.a. 23-224) that contains a V-type and a C2-type Ig-like domain (a.a. 27-112 and 128-203), followed by a single transmembrane domain (a.a. 225-245) and a cytoplasmic tail (a.a. 246-333). CRACC cytoplasmic domain contains one immunoreceptor tyrosine-based switch motif (ITSM; 278-TFYSTV-283, numbering based on mature murine sequence), which upon phosphorylation (pY300) recruits SAP family adaptor EWS-Fli1-activated transcript-2 (EAT-2) for signaling transduction. CRACC can also be phosphorylated on Tyr280. Unlike pY300, pY280 mediates the inhibitory function of CRACC in NK cells. In addition to NK cells, evidence indicates that CRACC is also involved in regulating B- and T-cell functions.

Specificity

Clone 4G2 reacted with murine T lymphoma BI-141 transfectants expressing mouse CRACC but not with those transfected with other members of the SLAM family (CD84, 2B4, Ly9, Ly108 or SLAM) (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305). Expected to react with all four spliced isoforms of murine CRACC reported by UniProt (Q8BHK6).

Immunogen

Epitope: Extracellular domain.
Human IgG1 Fc -tagged recombinant mouse CRACC extracellular domain.

Application

Anti-CRACC Antibody, clone 4G2 is an antibody against CRACC for use in Flow Cytometry, Affects Function, Immunoprecipitation.
Flow Cytometry Analysis: A representative lot immunostained murine T lymphoma BI-141 transfectants expressing mouse CRACC but not those transfected with other members of the SLAM family (CD84, 2B4, Ly9, Ly108 or SLAM) (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305).
Flow Cytometry Analysis: A representative lot detected CRACC expression on the surface of many types of mouse immune system cells, including NK cells, NKT cells, B cells, DCs, macrophages and activated T cells (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305).
Affects Function: A representative lot, in combination with a rabbit anti-rat IgG, cross-linked cell surface CRACC and induced similar CRACC phosphorylation in IL-2-activated NK cells from wild-type, SAP-deficient, or EAT-2–ERT–double knock (DKO) mice (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305).
Immunoprecipitation Analysis: A representative lot, in combination with a rabbit anti-rat IgG and protein A, immunoprecipitated CRACC from lysates of IL-2-activated mouse NK cells (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305).
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by Flow Cytometry in mouse splenocytes.

Flow Cytometry Analysis: 0.1 µg of this antibody detected CRACC-positive mouse splenocytes.

Target description

34.77 kDa (isoform 1), 34.87 kDa (isoform 2), 30.71 kDa (isoform 3), and 30.61 kDa (isoform 4) calculated.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG1κ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service