Skip to Content
Merck
All Photos(1)

Key Documents

MABF3060

Sigma-Aldrich

Anti-MERS-CoV RBD Antibody, clone m336

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$300.00
100 μG
$753.00

$300.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$300.00
100 μG
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$300.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

human

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

m336, monoclonal

mol wt

calculated mol wt 149.41 kDa

purified by

using Protein A

species reactivity

SARS coronavirus

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
neutralization: suitable

isotype

IgG1

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

vaccinia virus ... s> S(37616432)

General description

Spike glycoprotein (UniProt: K9N5Q8; also known as S glycoprotein, E2, Peplomer protein) is encoded by the S (also known as 3) gene (Gene ID: 14254594) in Middle East respiratory syndrome-related coronavirus (MERS-CoV). MERS-CoV is a positive-strand RNA virus of the betacoronavirus genus within the Coronaviridae family that causes severe respiratory syndrome in human. The mature MERS-CoV contains 4 structural proteins: Envelope (E), Membrane (M), Nucleocapsid (N), and the Spike protein (S). Spike protein is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off to generate the mature form that can be cleaved into three subunits known as S1 (aa 18-751), S2 (aa 752-1353), and S2 (aa 888-163) by host cell furin or other cellular proteases. Spike protein has an extracellular domain (aa 18-1296), a transmembrane domain (aa 1297-1317), and a cytoplasmic domain (aa 1318-1353). S1 subunit attaches the virion to the cell membrane by interacting with host receptor, initiating the infection. It interacts with host DPP4 to mediate viral entry. The S2 subunit mediates fusion of the virion and cellular membranes by acting as a class I viral fusion protein. Clone m336 binds to the receptor binding domain of spike protein that is localized in amino acids 360-601. It is shown to neutralized both pseudotyped and live MERS-CoV with high potency likely by competing with DPP4 for binding to the S glycoprotein. It does not exhibit reactivity with SARS-CoV2 spike protein. (Ref.: Ying, T., et al. (2014). J. Virol. 88(14); 7796-7805).

Specificity

Clone m336 is a recombinant human monoclonal antibody that detects spike protein in Middle East respiratory syndrome-related coronavirus (MERS-CoV). It targets an epitope within the receptor binding domain.

Immunogen

Generated with a large phage display library constructed by using cDNA of peripheral blood mononuclear cells (PBMC) from healthy volunteers and then panned using the RBD (360-601) of MERS-CoV.

Application

Quality Control Testing

Evaluated by ELISA with recombinant receptor binding domain (RBD) from spike protein of MERS-CoV.

ELISA Analysis (ELISA): Various dilutions of this antibody detected recombinant receptor binding domain (RBD) from spike protein of MERS-CoV.

Tested Applications

Neutralizing: A representative lot neutralized MERS-CoV RBD in Neutralizing applications (Ying, T. et al. (2014). J Virol.;88(14):7796-805; Folegatti, P.M., et al. (2020). Lancet Infect Dis.;20(7):816-826).

ELISA Analysis: A representative lot detected MERS-CoV RBD in ELISA applications (Ying, T. et al. (2014). J Virol.;88(14):7796-805; Folegatti, P.M., et al. (2020). Lancet Infect Dis.;20(7):816-826).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MERS-CoV RBD, clone m336, Cat. No. MABF3060, is a human recombinant monoclonal antibody that detects of Spike glycoprotein of Middle East respiratory syndrome-related coronavirus and is tested for use in ELISA and Neutralizing.

Physical form

Purified human recombinant monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service