Skip to Content
Merck
All Photos(1)

Key Documents

MABF2804

Sigma-Aldrich

Anti-Reticular fibroblasts Antibody, clone ER-TR7

Synonym(s):

Fibroblast marker

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$237.00
100 μG
$596.00

$237.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1048

Request a Bulk Order

Select a Size

Change View
25 μG
$237.00
100 μG
$596.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$237.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1048

Request a Bulk Order

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

ER-TR7, monoclonal

purified by

using protein G

species reactivity

mouse, human

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2a

epitope sequence

Unknown

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

General description

Connective tissue is responsible for maintaining the form of the body and its organs and provides cohesion and internal support. It includes several types of fibrous tissue that vary only in their density and cellularity. The cells of connective tissue include fibroblasts and adipose cells that are relatively stationary and several types of motile migrating cells, such as mast cells, macrophages, monocytes, lymphocytes, plasma cells, and eosinophils. These cells of connective tissue develop during embryonic life from the mesenchyme. Fibroblasts are the least specialized cells in the connective-tissue family. They are dispersed in connective tissue throughout the body, where they secrete a nonrigid extracellular matrix that is rich in type I and/or type III collagen. In skin fibroblasts originate from two distinct lineage of cells: one lineage populates the papillary dermis, and the other lineage populates the reticular dermis, that have distinct cell markers and distinct functional abilities. Clone ER-TR specifically detects an antigen present in, and produced by, reticular fibroblasts. This clone is useful to stain the microanatomy of various organs, in particular the connective tissue framework in lymphoid organs. It is also shown to detect reticular fibroblasts in thymus. (Ref.: Van Vliet, E., et al. (1984). Eur. J. Immunol. 14(6); 524-529).

Specificity

Clone ER-TR7 is a rat monoclonal antibody that detects Reticular fibroblasts.

Immunogen

Thymic stromal cells isolated from C3H mice.

Application

Quality Control Testing

Evaluated by Immunohistochemistry in Mouse thymus tissue sections.

Immunohistochemistry Applications: A 1:50 dilution of this antibody detected reticular fibroblasts in frozen section of Mouse thymus tissue.

Tested applications

Immunofluorescence Analysis: A representative lot detected Reticular fibroblasts clone ER-TR7 in Immunofluorescence applications (Van Vliet, E., et al. (1984). Eur J Immunol. 14(6):524-9; Nolte, M.A., et al. (2003). J Exp Med. 198(3):505-12; Sixt, M., et al. (2005). Immunity. 22(1):19-29; Link, A., et al. (2007). Nat Immunol. 8(11):1255-65; Odaka, C., et al. (2009). J Histochem Cytochem. 57(4):373-82; Schiavinato, A., et al. (2021). Eur J Immunol. 51(9):2345-2347).

Immunohistochemistry Applications: A representative lot detected Reticular fibroblasts clone ER-TR7 in Immunohistochemistry applications (Van Vliet, E., et al. (1984). Eur J Immunol. 14(6):524-9; Nolte, M.A., et al. (2003). J Exp Med. 198(3):505-12; Sixt, M., et al. (2005). Immunity. 22(1):19-29; Link, A., et al. (2007). Nat Immunol.;8(11):1255-65; Odaka, C., et al. (2009). J Histochem Cytochem. 57(4):373-82; Schiavinato, A., et al. (2021). Eur J Immunol. 51(9):2345-2347).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Reticular fibroblasts, clone ER-TR7, Cat. No. MABF2804, is a rat monoclonal antibody that detects Reticular fibroblasts and is tested for use in Immunofluorescence and Immunohistochemistry.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service