Skip to Content
Merck
All Photos(1)

Key Documents

MABF28

Sigma-Aldrich

Anti-PAR-3 Antibody, clone 8E8

clone 8E8, from mouse

Synonym(s):

Proteinase-activated receptor 3, Coagulation factor II receptor-like 2, Thrombin receptor-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$632.00

$632.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8E8, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

activity assay: suitable
flow cytometry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PARD3(56288)

General description

PAR-3 (also known as Coagulation factor II receptor-like 2, Thrombin receptor-like 2 or F2RL2) is an adapter protein involved in asymmetrical cell division and cell polarization processes. It seems to play a central role in the formation of epithelial tight junctions. Its association with PARD6B may prevent the interaction of PAR-3 with F11R/JAM1, thereby preventing tight junction assembly. The PARD6-PAR-3 complex links GTP bound Rho small GTPases to atypical protein kinase C proteins. PAR-3 interacts with PARD6A and PARD6B. Isoform 2, but not at least isoform 3 interacts with PRKCZ. PAR-3 interacts with PRCKI. PAR-3 forms part of a complex with PARD6A or PARD6B, PRKCI or PRKCZ and CDC42 or RAC1. PAR-3 interacts with F11R/JAM1. Antibodies against PAR-3 are present in sera from patients with cutaneous T cell lymphomas.

Immunogen

KLH-conjugated linear peptide corresponding to human PAR-3.

Application

Anti-PAR-3 Antibody, clone 8E8 detects level of PAR-3 & has been published & validated for use in FC, EA.
Flow Cytometry Analysis: A previous lot was used by an independent laboratory in FC. (Petrova, Y., et al. (2008). Centr Eur J Immunol. 33 (1):14-18.)

Activity Assay Analysis (platelet aggregation): A previous lot was used by an independent laboratory in platelet aggregation assay. (Petrova, Y., et al. (2008). Centr Eur J Immunol. 33 (1):14-18.)
Research Category
Infectious Diseases
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

Evaluated by Flow Cytometry in mouse platelets from washed whole blood (heparinized).

Flow Cytometry Analysis: 2 µg of this antibody detected PAR-3 in mouse platelets from washed whole blood (heparinized).

Target description

40 kDa calculated

Linkage

Replaces: MABS174

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse platelets from washed whole blood (heparinized)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Nakajima et al.
Journal of cell science, 123(Pt 4), 555-566 (2010-01-28)
Cell-shape change in epithelial structures is fundamental to animal morphogenesis. Recent studies identified myosin-II as the major generator of driving forces for cell-shape changes during morphogenesis. Lulu (Epb41l5) is a major regulator of morphogenesis, although the downstream molecular and cellular
Michael L Drummond et al.
The Journal of cell biology, 217(9), 3255-3266 (2018-06-28)
Primary cilia are polarized organelles that allow detection of extracellular signals such as Hedgehog (Hh). How the cytoskeleton supporting the cilium generates and maintains a structure that finely tunes cellular response remains unclear. Here, we find that regulation of actin
Kaviya Chinnappa et al.
Science advances, 8(2), eabj4010-eabj4010 (2022-01-13)
The evolutionary expansion and folding of the mammalian cerebral cortex resulted from amplification of progenitor cells during embryonic development. This process was reversed in the rodent lineage after splitting from primates, leading to smaller and smooth brains. Genetic mechanisms underlying
Virginia Fernández et al.
The EMBO journal, 39(21), e105479-e105479 (2020-09-29)
Structural integrity and cellular homeostasis of the embryonic stem cell niche are critical for normal tissue development. In the telencephalic neuroepithelium, this is controlled in part by cell adhesion molecules and regulators of progenitor cell lineage, but the specific orchestration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service