Skip to Content
Merck
All Photos(2)

Key Documents

MABF2781

Sigma-Aldrich

Anti-LCMV Nucleoprotein Antibody, clone VL-4

Synonym(s):

LCMV Nucleocapsid Protein, Protein N

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$233.00
100 μL
$590.00

$233.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$233.00
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$233.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

VL-4, monoclonal

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
activity assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
neutralization: suitable

isotype

IgG2aκ

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

vaccinia virus ... N(956592)

Specificity

Clone VL-4 is a rat monoclonal antibody that detects nucleoprotein of Lymphocytic choriomeningitis virus (LCMV).

Immunogen

Lymphocytic choriomeningitis virus (LCMV).

Application

Quality Control Testing

Evaluated by ELISA with recombinant LCMV nucleoprotein .

ELISA Analysis: A serial dilution of this antibody detected recombinant CLMV nucleoprotein.

Tested Applications

ELISA Analysis: A representative lot detected LCMV Nucleoprotein in ELISA application (Eschli, B., et al. (2007). J Virol. 81(21):11650-7).

Flow Cytometry Analysis: A representative lot detected LCMV Nucleoprotein in Flow Cytometry application (Herz, J., et al. (2015). J Exp Med. 212(8):1153-69).

Neutralizing: A representative lot detected LCMV Nucleoprotein in Neutralizing applications (Eschli, B., et al. (2007). J Virol. 81(21):11650-7).

Activity Assay: A representative lot detected Nucleoprotein in lymphocytic choriomeningitis virus (LCMV) (Data courtesy of the Division of Experimental Virology, Department of Biomedicine, University of Basel, Switzerland).

Immunohistochemistry Applications: A representative lot detected LCMV Nucleoprotein in Immunohistochemistry applications (Herz, J., et al. (2015). J Exp Med. 212(8):1153-69; Urata, S., et al. (2018). PLoS Pathog. 14(7):e1007172).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Nucleoprotein (UniProt: P07400; also known as Nucleocapsid protein, Protein N) is encoded by the N gene (Gene ID: 127897) in Lymphocytic choriomeningitis virus (LCMV). LCMV is a murine enveloped RNA virus of the arenavirus family. Its viral surface is densely covered with viral fusion glycoprotein spikes that form heterodimeric trimers, consisting of the peripheral receptor binding glycoprotein 1 (GP-1) and a transmembrane subunit (GP-2). The large segment of LCMV genome encodes for the RNA-dependent-RNA-polymerase (L) and the matrix protein (Z) that mediates viral assembly and budding. Its small segment (S) encodes the glycoprotein precursor (GPC) and the viral nucleoprotein (NP). GPC is then co-translationally cleaved by signal peptidase to produce a stable 58 amino acid stable signal peptide (SSP) and GPC that is post-translationally processed by the cellular site 1 protease to form GP-1 and GP-2. GP-1 is reported to mediate virion attachment to the cell surface followed by cell entry via receptor-mediated endocytosis. The GP-2 subunit is responsible for the pH-dependent fusion event in the acidic environment of the endosome to complete the virus cell entry process and release of virus ribonucleoprotein into the cell cytoplasm to initiate transcription and replication of the viral genome. The nucleoprotein of LCMV encapsidates the genome and protects it from nucleases. The encapsidated genomic RNA known as the nucleocapsid (NC) serves as template for viral transcription and replication. Clone VL-4 specifically reacts with LCMV nucleoprotein and does not exhibit reactivity with vaccinia or influenza virus infected cells. (Ref.: Urata, S., et al. (2018). PLoS Pathog 14(7): e1007172; Eschli, B., et al. (2007). J. Virol. 81(21); 11650-11657).

Physical form

Purified rat monoclonal antibody IgG2a in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service