Skip to Content
Merck
All Photos(1)

Key Documents

MABF2761

Sigma-Aldrich

Anti-CXCL1/GROa Antibody, clone HL2401

Synonym(s):

C-X-C motif chemokine 1;GRO-alpha(1-73);Melanoma growth stimulatory activity;MGSA;Neutrophil-activating protein 3;NAP-3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$237.00
100 μG
$596.00

$237.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$237.00
100 μG
$596.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$237.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

HL2401, monoclonal

mol wt

calculated mol wt 11.3 kDa
observed mol wt ~15 kDa

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CXCL1(2919)

General description

Growth-regulated alpha protein (UniProt: P09341; also known as C-X-C motif chemokine 1, CXCL1, GRO-alpha(1-73), Melanoma growth stimulatory activity, MGSA, Neutrophil-activating protein 3, NAP-3) is encoded by the CXCL1 (also known as GRO, GRO1, GROA, MGSA, SCYB1) gene (Gene ID: 2919) in human. CXCL1 is a multi-functional chemokine that displays chemotactic activity for neutrophils. It is synthesized with a signal peptide (aa 1-34), which is subsequently cleaved off to produce the mature protein that can be further cleaved into three chains (aa 4-73; 5-73; and 6-73) via proteolytic cleavage after secretion from peripheral blood monocytes. CXCL1 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. Its levels are reported to be low under basal conditions but increase significantly following active infection. It plays a role in recruiting and activating neutrophils for microbial killing at the tissue site and can also activate the release of proteases and reactive oxygen species. CXCL1 signals through binding to CXCR2, a G-protein coupled receptor. Aberrant expression of CXCL1 has been reported in several cancers and has been associated with oncogenesis, metastasis, angiogenesis and chemoresistance. Clone HL2401 is shown to specifically binds to CXCL1, although it may show slight binding to CXCL2. It is shown to reduce both cellular proliferation and invasion of cancer cells overexpressing CXCL1. It also induces apoptosis and impairs tumor vasculature. (Ref.: Miyake, M., et al. (2019). Theranostics.;9(3):853-867; Yang C., et al. (2019). Int. J. Oncol. 55(3); 684-696Wuyts, A., et al. (1999). Eur. J. Biochem. 260(2); 421-429).

Specificity

Clone HL2401 is a mouse monoclonal antibody that detects CXCL1.

Immunogen

Native human CXCL1 protein.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Human CXCL1 protein.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant Human CXCL1 protein.

Tested Applications

ELISA Analysis: A representative lot detected CXCL1 in ELISA application (Miyake, M., et al. (2019). Theranostics. 9(3):853-867).

Inhibition Assay: A representative lot inhibited proliferation and invasion of bladder and prostate cells and disrupted endothelial sprouting in vitro. (Miyake, M., et al. (2019). Theranostics. 9(3):853-867).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CXCL1 in Immunoprecipitation application (Miyake, M., et al. (2019). Theranostics. 9(3):853-867).

Western Blotting Analysis: A representative lot detected CXCL1 in Western Blotting application (Miyake, M., et al. (2019). Theranostics. 9(3):853-867).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.02% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service