Skip to Content
Merck
All Photos(3)

Key Documents

MABF248

Sigma-Aldrich

Anti-SLFN11 Antibody, clone 4G9

clone 4G9, 0.25 mg/mL, from mouse

Synonym(s):

Schlafen family member 11, SLFN11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$632.00

$632.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1405

Request a Bulk Order

Select a Size

Change View
100 μL
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1405

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G9, monoclonal

species reactivity

human

concentration

0.25 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLFN11(91607)

General description

SLFN11, also known as Schlafen family member 11, and encoded by the gene SLFN11, is an interesting protein that is part of our innate defense against retroviruses. The protein is induced by interferon and acts to inhibit the translation of retrovirus specific proteins. SLFN11 also plays a role in cell cycle arrest and apoptosis induction, particularly in response to DNA damage. SLFN11 is localized to the nucleus and is widely expressed and up regulated in various cancers including ovarian and adenocarcinomas, and recent work suggests that SLFN11 may behave as a biomarker for prediction of response to DNA-damaging agents in the clinical setting.

Immunogen

Recombinant protein corresponding to human SLFN11.

Application

Anti-SLFN11 Antibody, clone 4G9 is an antibody against SLFN11 for use in western blotting & IP.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Western Blotting/Immunoprecipitation Analysis: A representative lot immunoprecipitated SLFN11 from transfected HEK293 cell lysates. The same lot was used to detect SLFN11 in Western Blotting (Prof. Michael David, University of California, San Diego.).

Electron Mobility Shift Assay Analysis: A representative lot from an independent laboratory detected SLFN11 bound t-RNA (Li, M., et al. (2012). Nature. 491(7422):125-128.).

Quality

Evaluated by Western Blotting in HEK293T cell lysates transfected with SLFN11 or a negative control.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SFN11 in HEK293T cell lysates transfected with SLFN11 and not in HEK293T cell lysates transfected with a negative control.

Target description

~100 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marketa Bednarikova et al.
Cancers, 14(10) (2022-05-29)
Platinum-based chemotherapy has been the cornerstone of systemic treatment in ovarian cancer. Since no validated molecular predictive markers have been identified yet, the response to platinum-based chemotherapy has been evaluated clinically, based on platinum-free interval. The new promising marker Schlafen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service