Skip to Content
Merck
All Photos(1)

Key Documents

MABF2336

Sigma-Aldrich

Anti-HCV E2 Antibody, clone AR3A

clone AR3A

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$300.00
100 μL
$765.00

$300.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$300.00
100 μL
$765.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$300.00


Estimated to ship on01 June 2025


Request a Bulk Order

clone

AR3A, recombinant monoclonal

Quality Level

isotype

IgG1

Specificity

Clone AR3A is a human recombinant antibody produced from Fab fragment C1, expressed in E.coli and converted into full-length IgG1 in CHO-K1 cells. It detects E2 envelope protein in Hepatitis C virus.

Immunogen

Antibody Fab fragment C1 expressed in E. coli, purified, and converted into full-length IgG1 molecule in CHO-K1 cells.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service