Skip to Content
Merck
All Photos(2)

Key Documents

MABF2299

Sigma-Aldrich

Anti-IL-1α Antibody, clone 7E5.14.5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$300.00
100 μG
$753.00

$300.00


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$300.00
100 μG
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$300.00


Estimated to ship on22 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

7E5.14.5, monoclonal

mol wt

calculated mol wt 30.61 kDa
observed mol wt ~18 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

General description

Interleukin-1 alpha (UniProt: P01583; also known as IL-1 alpha, Hematopoietin-1) is encoded by the IL1A (also known as IL1F1) gene (Gene ID: 3552) in human. IL-1a is constitutively expressed in several healthy tissues in a steady state and its expression can be increased in response to a number of stimuli. Absolute amounts of IL-1a vary among cell types, but barrier cells, such as endothelial and epithelial cells, express higher amounts at steady state. IL-1a is synthesized with a propeptide (aa 1-112) and is post-translationally modified by phosphorylation at serine 90, myristoylation on lysine 82, and acetylation on lysine 82. After synthesis, pro-IL-1a is localized into the nucleus, cytosol or lysosomal compartment or is displayed on the outer leaflet of the plasma membrane. Its intracellular localization can change in response to specific stimuli. Pro-IL-1a can also be cleaved by calpain to generate IL-1 -N-terminal domain (NTP) and a mature C-terminal IL-1a form. The C-terminal domain is catalytically active and is capable of signaling via IL-1R. Both IL-1a -NTP and pro-IL-1a contains a functional NLS signal and bind HAX1 to allow translocation into the nucleus. In the nucleus, IL-1a -NTP binds transcription regulation factors and activates expression of proinflammatory cytokines and chemokines independently of IL-1R1 signaling. The biological functions of IL-1a also involve stimulation of thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. (Ref.: Malik, A., and Kanneganti, TD. (2018). Immunol. Rev. 281(1); 124-137; Di Paolo, NC., and Shayakhmetov, DM (2016). Nat. Commun. 17(8); 906-913).

Specificity

Clone 7E5.14.5 is a mouse monoclonal antibody that detects human IL-1a .

Immunogen

Full-length recombinant human IL-1alpha (without the propeptide).

Application

Quality Control Testing

Evaluated by Western Blotting with human recombinant IL-1a.

Western Blotting Analysis: A 1,000 dilution of this antibody detected human recombinant IL-1a.

Tested Applications


Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected IL-1a in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IL-1a, clone 7E5.14.5, Cat. No. MABF2299, is a mouse monoclonal antibody that detects Interluekin-1a and is is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service