Skip to Content
Merck
All Photos(1)

Key Documents

MABF223

Sigma-Aldrich

Anti-TMEM173/STING Antibody, clone 1B1.1

clone 1B1.1, from mouse

Synonym(s):

Stimulator of interferon genes protein, hSTING, Endoplasmic reticulum interferon stimulator, ERIS, Mediator of IRF3 activation, hMITA, Transmembrane protein 173

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$554.00

$554.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$554.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$554.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B1.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

TMEM173/STING, also known as STING, or Endoplasmic reticulum interferon stimulator, ERIS, Mediator of IRF3 activation, MMITA or Transmembrane protein 173, and encoded by the gene TMEM173/ERIS/MITA/STING, is a facilitator of innate immune signaling that acts as a sensor of cytosolic DNA from bacteria and viruses and promotes the production of type I interferons. STING acts by recognizing and binding cyclic di-GMP (c-di-GMP), a second messenger produced by bacteria, and cyclic GMP-AMP (cGAMP), a messenger produced in response to DNA virus in the cytosol. Upon binding STING is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon and exert a potent anti-viral state. STING also mediates cell death signaling via the activation of the ERK pathway. STING is found in multiple intracellular locations including the endoplasmic reticulum, plasma cell membrane, mitochondrion outer membrane, and perinuclear regions. STING is expressed in the B-cell lineage both immature and mature B cells but not plasma cells. Mutations in STING cause defects in innate immunity and compromised immune health.

Immunogen

GST-tagged recombinant protein corresponding to human TMEM173/STING.

Application

Anti-TMEM173/STING Antibody, clone 1B1.1 | MABF223 is an antibody against TMEM173/STING Antibody for use in Western Blotting.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected TMEM173/STING in 10 µg of Jurkat cell lysate.

Target description

~37 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service