Skip to Content
Merck
All Photos(1)

Key Documents

MABF216

Sigma-Aldrich

Anti-Microsialin/CD68 Antibody, clone 9

clone 9, 1 mg/mL, from rat

Synonym(s):

Macrosialin, CD68

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$454.00

$454.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$454.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$454.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9, monoclonal

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd68(12514)

General description

Microsialin/CD68, also known as Macrosialin, Gp110, CD_antigen 68, or CD68, and encoded by the gene CD68, is a membrane protein expressed in macrophages (and other cells) that appears to play a role in the phagocytic activities of tissue macrophages both in intracellular lysosomal metabolism but also in extracellular cell to cell interactions and cell pathogen interactions. Microsialin binds to tissue and organ-specific lectins or selectins mediating homing of macrophage subsets to particular sites. Also, rapid recirculation of Microsialin from the endosomes and lysosomes to the plasma membrane may allow macrophages to crawl over selection-bearing substrates or other cells. Microsialin is expressed in blood monocytes and tissue macrophages primarily but is also found expressed in lymphocytes, fibroblasts and endothelial cells so it is not an exclusive marker for macrophages. Also Microsialin is expressed on many tumor cells which seem to allow the tumor cells to attach to selectins on the vascular endothelium facilitating their dissemination to secondary sites.

Immunogen

Recombinant protein corresponding to mouse Microsialin/CD68.

Application

Anti-Microsialin/CD68 Antibody, clone 9 is an antibody against Microsialin/CD68 for use in flow cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service