Skip to Content
Merck
All Photos(2)

Key Documents

MABF2125

Sigma-Aldrich

Anti-Properdin Antibody, clone 6E9E6

clone 6E9E6, from mouse

Synonym(s):

Complement factor P

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$189.00
100 μL
$481.00

$189.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$189.00
100 μL
$481.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$189.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

6E9E6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CFP(5199)

General description

Properdin (UniProt: P27918; also known as Complement factor P) is encoded by the CEP (also known as PFC) gene (Gene ID: 5199) in human. (ABF185: Properdin (UniProt P27918; also known as Complement factor P) is encoded by the CFP (also known as PFC, PFD) gene (Gene ID 5199) in human. Properdin is a plasma glycoprotein and the only known positive regulator of the complement cascade. It binds and stabilizes the alternative pathway (AP) C3 convertase C3bBb and may also serve as a platform to form new C3bBb convertases on cell surface. Properdin-mediated AP activation plays an important role in host defense, and properdin deficiency (PFD) increases the risk for Neisseria meningitidis infection in human. Properdin-deficiency exacerbated disease condition in a murine model of factor H- (fH-) related C3 glomerulopathy, but prevented the development of allergen-induced airway inflammation. Human properdin is produced with a signal peptide sequence (a.a. 1-27), which is removed posttranslationally to yield the mature protein (a.a. 28-469) composed entirely of seven TSP type-1 domains (domain 0 through 6; a.a. 28-462). TSP type-1 domains 0 and 6 bind to each other and mediate multimerization.

Specificity

Clone 6E9E6 specifically detects human Properdin.

Immunogen

Purified properdin from human plasma.

Application

Anti-Properdin, clone 6E9E6 Antibody, Cat. No. MABF2125, is a mouse monoclonal antibody that detects Properdin and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected Properdin in human tonsil tissue sections.
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting with purified human properdin.

Western Blotting Analysis: A 1:500 dilution of this antibody detected purified human properdin.

Target description

~51 kDa observed; 51.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service