Skip to Content
Merck
All Photos(1)

Key Documents

MABF2087

Sigma-Aldrich

Anti-Adenylate Cyclase Toxin Antibody, B. pertussis Antibody, clone 9D4

clone 9D4, from mouse

Synonym(s):

Adenylate cyclase toxin, Bordetella pertussis Toxin, ACT

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$252.00
100 μL
$641.00

$252.00


Estimated to ship on14 April 2025



Select a Size

Change View
25 μL
$252.00
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$252.00


Estimated to ship on14 April 2025


biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9D4, monoclonal

species reactivity

bacteria

packaging

antibody small pack of 25 μL

technique(s)

dot blot: suitable
western blot: suitable

isotype

IgG2aλ

target post-translational modification

unmodified

General description

Adenylate Cyclase Toxin (UniProt: C8C508; also known as ACT or Cya A) is one of several virulence factors produced by the bacterium Bordetella pertussis. Bordetella pertussis, a gram-negative bacterium, is shown to be the causative agent for whooping cough. It produces several virulence factors, including Cya A. The CyaA toxin is a bifunctional protein of 1,706 residues consisting of an NH2-terminal catalytic domain of 400 amino acids and a COOH-terminal part with 1,306 residues. The CyaA polypeptide is synthesized as an inactive protoxin, which is then converted to an active toxin by posttranslational palmitoylation of two internal lysine residues (Lys 856 and 963). Upon interaction of its C-terminal hemolysin moiety with the cell surface receptor alphaM-beta2 integrin ((Cd11b/Cd18), the N-terminal cyclase domain translocates into the host cell cytosol where it generates supraphysiological levels of cAMP concentration, which inhibits host cell anti-bacterial activities and apoptotic death of macrophages. Cya A toxin is also shown to elicit potassium ion efflux from erythrocytes in a process that is thought to be a precursor event to osmotic lysis of erythrocytes. Clone 9D4 binds to an epitope of AC toxin in the glycine- and aspartate- rich region. Hence, it recognizes other RTX toxins in addition to AC toxin. Clone 9D4 increases the fraction of AC toxin (CyaA) that is secreted to cell surface and release of CyaA into culture supernatant(Ref.: Guermonprez, P., et al. (2001). J. Exp. Med. 193(9); 1035-1044; Gray, MC., et al. (2001). J. Bacteriol. 183(20); 5904-5910; Holubova, J., et al. (2012). Infect. Immun. 80(3); 1181-1192).

Specificity

Clone 9D4 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis.

Immunogen

Adenylate cyclase toxin from Bordetella pertussis.

Application

Anti-Adenylate Cyclase Toxin, B. pertussis, clone 9D4, Cat. No. MABF2087 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis and has been tested for use in Dot Blot, Function Assay and Western Blotting.
Function Analysis: A representative lot increased the fraction of AC toxin (CyaA) that is secreted to cell surface and release of CyaA into culture supernatant. (Gray, M.C., et. al. (2001). J Bacteriol. 183(20):5904-10).

Western Blotting Analysis: A representative lot detected Adenylate Cyclase Toxin, B. pertussis in Western Blotting applications (Holubova, J., et. al. (2012). Infect Immun. 80(3):1181-92; Lee, S.J., et. al. (1999). Infect Immun. 67(5):2090-5).

Dot Blot Analysis: A representative lot detected Adenylate Cyclase Toxin, B. pertussis in Dot Blot applications (Lee, S.J., et. al. (1999). Infect Immun. 67(5):2090-5).



Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in Adenylate cyclase toxin from Bordetella pertussis.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Adenylate Cyclase Toxin from from Bordetella pertussis.

Target description

~220 kDa observed.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service