Skip to Content
Merck
All Photos(1)

Key Documents

MABF1986M

Sigma-Aldrich

Anti-NKR-P1G Antibody, clone ADA3

clone ADA3, from hamster(Armenian)

Synonym(s):

Killer cell lectin-like receptor subfamily B member 1, Immunoreceptor NKR-P1E, Killer cell lectin-like receptor subfamily B member 1G, Natural killer cell surface protein NKR-P1G, Natural killer lectin-like receptor 1E

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$189.00
200 μL
$452.00

$189.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$189.00
200 μL
$452.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$189.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

hamster (Armenian)

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ADA3, monoclonal

species reactivity

rat

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Killer cell lectin-like receptor subfamily B member 1 (UniProt: Q0ZUP0; also known as Immunoreceptor NKR-P1E, Killer cell lectin-like receptor subfamily B member 1G, Natural killer cell surface protein NKR-P1G, Natural killer lectin-like receptor 1E) is encoded by the Klrb1 (also known as Klrb1d, Klrb1g, Klrb6, Nkrp1e, Nkrp1g) gene (Gene ID: 689817) in rat. NKR-P1G is a single-pass type II membrane protein with a cytoplasmic domain (aa 1-42), a helical domain (aa 43-63), and an extracellular domain (aa 64-214). NKR-P1G is expressed by subsets of lymphocytes with highest prevalence in gut and liver mucosa. The number of NKR-P1G+ cells are shown to increase with age and its expression level is modulated by cytokines, with an increase induced by inflammatory cytokines. It is shown to be mainly expressed by Ly49s3+ cells. It is reported to inhibit cytotoxicity and calcium responses in IL-2-activated NK cells. NKR-P1G is a potent inhibitory receptor for a panel of Clr ligands. The intracellular domain of NKR-P1G contains an immunoreceptor tyrosine-based inhibitory motif that is phosphorylated by Src-kinases upon activation. This allows recruitment of phosphotyrosine phosphatases, which in turn negatively regulates the signaling cascades that is essential for cytotoxicity and cytokine production. (Ref.: Kveberg, L et al (2015). Eur J Immunol. 45(2); 501-512).

Specificity

Clone ADA3 is an Armenian hamster monoclonal antibody that detects Killer cell lectin-like receptor subfamily B member 1 (NKR-P1G) in rat. It targets an epitope within the extracellular domain.

Immunogen

Chinese hamster ovary (CHO) cells stably transfected with FLAG-tagged extracellular portion of NKR-P1G.
Epitope: extracellular domain

Application

Anti-NKR-P1G, clone ADA3, Cat. No. MABF1986 is an Armenian hamster monoclonal antibody that detects NKG-P1G and has beent tested for use in Flow Cytometry and Immunoprecipitation.
Immunoprecipitation Analysis: A representative lot detected NKR-PIG in Immunoprecipitation applications (Kveberg, L., et. al. (2015). Eur J Immunol. 45(2):501-12).

Flow Cytometry Analysis: A representative lot detected NKR-PIG in Flow Cytometry applications (Kveberg, L., et. al. (2015). Eur J Immunol. 45(2):501-12).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in Ltk (mouse fibroblast) cells stably transfected with FLAG-tagged rat NKR-P1G.

Flow Cytometry Analysis: 1 µg of this antibody detected NKR-P1G in Ltk (mouse fibroblast) cells stably transfected with FLAG-tagged rat NKR-P1G.

Target description

23.96 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified Armenian hamster monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service