Skip to Content
Merck
All Photos(1)

Key Documents

MABF176E

Sigma-Aldrich

Anti-CD2 (human), PerCP, clone RPA-2.10 Antibody

clone RPA-2.10, from mouse, Peridinin-Chlorophyll-Protein Complex

Synonym(s):

T-cell surface antigen CD2, Erythrocyte receptor, LFA-2, LFA-3 receptor, Rosette receptor, T-cell surface antigen T11/Leu-5, CD antigen CD2, CD2

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
$227.00
100 TESTS
$559.00
500 TESTS
$2,480.00

$227.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
25 TESTS
$227.00
100 TESTS
$559.00
500 TESTS
$2,480.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$227.00


Estimated to ship on10 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

Peridinin-Chlorophyll-Protein Complex

antibody form

purified antibody

antibody product type

primary antibodies

clone

RPA-2.10, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD2(914)

Related Categories

General description

The RPA-2.10 antibody reacts with human CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD58 in the human and is expressed on the cell surface of 80-90% of human peripheral blood lymphocytes, a subset of NK cells, and all mature T cells. CD2 mediates lymphocyte adhesion and is involved in T cell activation.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Anti-CD2 (human), PerCP, clone RPA-2.10 Antibody is an antibody against CD2 for use in Flow Cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected CD2 in one million human peripheral blood lymphocytes.

Physical form

Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service